Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139392826-139393057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROB1
Entrez Gene ID
389333
Gene Name
proline rich basic protein 1
Gene Aliases
C5orf65
UniGene
Hs.729482
Gene Chromosome Location
Chr.5: 139391946-139395196 on Build GRCh38
Transcripts
NM_001161546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4842976 c.2255G>A p.R752Q Chr.5: 139392827-139392827 on Build GRCh38
4827688 c.2049C>T p.F683F Chr.5: 139393033-139393033 on Build GRCh38
5799984 c.2062C>G p.L688V Chr.5: 139393020-139393020 on Build GRCh38
3749904 c.2238G>A p.E746E Chr.5: 139392844-139392844 on Build GRCh38
6347879 c.2037C>A p.Y679* Chr.5: 139393045-139393045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035763371 Chr.5: 139392932-139392932 on Build GRCh38
rs745551955 Chr.5: 139392926-139392926 on Build GRCh38
rs988336858 Chr.5: 139393011-139393011 on Build GRCh38
rs1001568333 Chr.5: 139392929-139392929 on Build GRCh38
rs770044205 Chr.5: 139393002-139393002 on Build GRCh38
rs368156469 Chr.5: 139392910-139392910 on Build GRCh38
rs559526824 Chr.5: 139392983-139392983 on Build GRCh38
rs746858794 Chr.5: 139392998-139392998 on Build GRCh38
rs547634868 Chr.5: 139392882-139392882 on Build GRCh38
rs779933737 Chr.5: 139392958-139392958 on Build GRCh38
rs1045005105 Chr.5: 139392903-139392903 on Build GRCh38
rs978980918 Chr.5: 139393015-139393015 on Build GRCh38
rs1010338530 Chr.5: 139392991-139392991 on Build GRCh38
rs776611510 Chr.5: 139393030-139393030 on Build GRCh38
rs946487506 Chr.5: 139393016-139393016 on Build GRCh38
rs962137479 Chr.5: 139392896-139392896 on Build GRCh38
rs10063870 Chr.5: 139392844-139392844 on Build GRCh38
rs891014103 Chr.5: 139392883-139392883 on Build GRCh38
rs758102302 Chr.5: 139392944-139392944 on Build GRCh38
rs968916692 Chr.5: 139393014-139393014 on Build GRCh38
rs928225780 Chr.5: 139393001-139393001 on Build GRCh38
rs895911645 Chr.5: 139392953-139392953 on Build GRCh38
rs778701207 Chr.5: 139392884-139392884 on Build GRCh38
rs1015360813 Chr.5: 139392893-139392893 on Build GRCh38
rs34131406 Chr.5: 139392901-139392901 on Build GRCh38
rs748366649 Chr.5: 139392993-139392993 on Build GRCh38

View Full Product Details