Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75617073-75617334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAT1
Entrez Gene ID
23536
Gene Name
adenosine deaminase, tRNA specific 1
Gene Aliases
HADAT1
UniGene
Hs.729312
Gene Chromosome Location
Chr.16: 75596879-75623323 on Build GRCh38
Transcripts
NM_001324444.1 NM_001324445.1 NM_001324446.1 NM_001324448.1 NM_001324449.1 NM_001324450.1 NM_001324451.1 NM_001324452.1 NM_001324453.1 NM_012091.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
973756 c.394A>G p.I132V Chr.16: 75617172-75617172 on Build GRCh38
4659804 c.309A>G p.Q103Q Chr.16: 75617257-75617257 on Build GRCh38
3512282 c.312C>T p.L104L Chr.16: 75617254-75617254 on Build GRCh38
6594722 c.366delA p.G123fs*49 Chr.16: 75617200-75617200 on Build GRCh38
4659803 c.414C>T p.S138S Chr.16: 75617152-75617152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951237537 Chr.16: 75617279-75617279 on Build GRCh38
rs556868899 Chr.16: 75617246-75617246 on Build GRCh38
rs771084797 Chr.16: 75617244-75617244 on Build GRCh38
rs762451594 Chr.16: 75617175-75617175 on Build GRCh38
rs766336731 Chr.16: 75617296-75617296 on Build GRCh38
rs765474487 Chr.16: 75617167-75617167 on Build GRCh38
rs763787303 Chr.16: 75617278-75617278 on Build GRCh38
rs746290396 Chr.16: 75617251-75617251 on Build GRCh38
rs1000155554 Chr.16: 75617273-75617273 on Build GRCh38
rs760834686 Chr.16: 75617141-75617141 on Build GRCh38
rs1042229018 Chr.16: 75617257-75617257 on Build GRCh38
rs745944115 Chr.16: 75617313-75617313 on Build GRCh38
rs771603941 Chr.16: 75617145-75617145 on Build GRCh38
rs903234096 Chr.16: 75617272-75617272 on Build GRCh38
rs778108393 Chr.16: 75617236-75617236 on Build GRCh38
rs910487418 Chr.16: 75617187-75617187 on Build GRCh38
rs750387911 Chr.16: 75617287-75617287 on Build GRCh38
rs200765442 Chr.16: 75617303-75617303 on Build GRCh38
rs901699156 Chr.16: 75617242-75617242 on Build GRCh38
rs139320152 Chr.16: 75617144-75617144 on Build GRCh38
rs758761615 Chr.16: 75617233-75617233 on Build GRCh38
rs754796582 Chr.16: 75617302-75617302 on Build GRCh38
rs113842565 Chr.16: 75617196-75617196 on Build GRCh38
rs773485691 Chr.16: 75617135-75617135 on Build GRCh38
rs369904477 Chr.16: 75617103-75617103 on Build GRCh38
rs765370296 Chr.16: 75617197-75617197 on Build GRCh38
rs751111918 Chr.16: 75617180-75617180 on Build GRCh38
rs766950900 Chr.16: 75617283-75617283 on Build GRCh38
rs752395808 Chr.16: 75617172-75617172 on Build GRCh38
rs753618243 Chr.16: 75617299-75617299 on Build GRCh38
rs866502602 Chr.16: 75617156-75617156 on Build GRCh38
rs750292116 Chr.16: 75617205-75617205 on Build GRCh38
rs770205207 Chr.16: 75617252-75617252 on Build GRCh38
rs144186738 Chr.16: 75617199-75617199 on Build GRCh38
rs373642560 Chr.16: 75617222-75617222 on Build GRCh38
rs757416690 Chr.16: 75617308-75617308 on Build GRCh38
rs781070872 Chr.16: 75617312-75617312 on Build GRCh38
rs749096982 Chr.16: 75617225-75617225 on Build GRCh38
rs756741764 Chr.16: 75617181-75617181 on Build GRCh38
rs779656369 Chr.16: 75617223-75617223 on Build GRCh38
rs375750022 Chr.16: 75617231-75617231 on Build GRCh38
rs777254859 Chr.16: 75617149-75617149 on Build GRCh38
rs775773103 Chr.16: 75617260-75617260 on Build GRCh38
rs149203549 Chr.16: 75617307-75617307 on Build GRCh38
rs1024085929 Chr.16: 75617137-75617137 on Build GRCh38
rs763369581 Chr.16: 75617265-75617265 on Build GRCh38
rs199978571 Chr.16: 75617240-75617240 on Build GRCh38
rs759967288 Chr.16: 75617150-75617150 on Build GRCh38
rs778977013 Chr.16: 75617264-75617264 on Build GRCh38
rs772206631 Chr.16: 75617126-75617126 on Build GRCh38
rs1032951904 Chr.16: 75617306-75617306 on Build GRCh38
rs756065049 Chr.16: 75617288-75617288 on Build GRCh38
rs773947279 Chr.16: 75617282-75617282 on Build GRCh38

View Full Product Details