Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 225987788-225988019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDE2
Entrez Gene ID
163859
Gene Name
SDE2 telomere maintenance homolog
Gene Aliases
C1orf55, dJ671D7.1
UniGene
Hs.520192
Gene Chromosome Location
Chr.1: 225982702-225999365 on Build GRCh38
Transcripts
NM_152608.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5418732 c.1043A>C p.E348A Chr.1: 225987951-225987951 on Build GRCh38
5845639 c.1073C>T p.A358V Chr.1: 225987957-225987957 on Build GRCh38
679370 c.1078G>T p.E360* Chr.1: 225987916-225987916 on Build GRCh38
1146153 c.1114G>T p.E372* Chr.1: 225987916-225987916 on Build GRCh38
1473455 c.1012G>C p.D338H Chr.1: 225987982-225987982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377323585 Chr.1: 225987881-225987881 on Build GRCh38
rs201203772 Chr.1: 225987938-225987938 on Build GRCh38
rs977519552 Chr.1: 225987897-225987897 on Build GRCh38
rs770116276 Chr.1: 225987910-225987910 on Build GRCh38
rs767196123 Chr.1: 225987959-225987960 on Build GRCh38
rs763038533 Chr.1: 225987918-225987918 on Build GRCh38
rs774629028 Chr.1: 225987818-225987818 on Build GRCh38
rs781659210 Chr.1: 225987894-225987894 on Build GRCh38
rs544500712 Chr.1: 225987950-225987950 on Build GRCh38
rs759216599 Chr.1: 225987945-225987946 on Build GRCh38
rs377596622 Chr.1: 225987931-225987931 on Build GRCh38
rs775722843 Chr.1: 225987916-225987916 on Build GRCh38
rs370711348 Chr.1: 225987932-225987932 on Build GRCh38
rs771734154 Chr.1: 225987977-225987977 on Build GRCh38
rs964395692 Chr.1: 225987835-225987835 on Build GRCh38
rs746282071 Chr.1: 225987899-225987899 on Build GRCh38
rs3795442 Chr.1: 225987843-225987843 on Build GRCh38
rs753546756 Chr.1: 225987942-225987942 on Build GRCh38
rs375173160 Chr.1: 225987898-225987898 on Build GRCh38
rs371110722 Chr.1: 225987892-225987892 on Build GRCh38
rs914994306 Chr.1: 225987864-225987864 on Build GRCh38
rs1047570445 Chr.1: 225987975-225987975 on Build GRCh38
rs768794158 Chr.1: 225987920-225987920 on Build GRCh38
rs766238967 Chr.1: 225987836-225987836 on Build GRCh38
rs778233921 Chr.1: 225987859-225987859 on Build GRCh38
rs774230206 Chr.1: 225987927-225987927 on Build GRCh38
rs899212362 Chr.1: 225987940-225987940 on Build GRCh38
rs752211409 Chr.1: 225987988-225987988 on Build GRCh38
rs760583329 Chr.1: 225987937-225987937 on Build GRCh38
rs746944950 Chr.1: 225987856-225987856 on Build GRCh38
rs773260083 Chr.1: 225987936-225987936 on Build GRCh38
rs768653970 Chr.1: 225987906-225987906 on Build GRCh38
rs773825914 Chr.1: 225987925-225987926 on Build GRCh38
rs753034351 Chr.1: 225987976-225987976 on Build GRCh38

View Full Product Details