Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95436781-95437052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE3
Entrez Gene ID
161176
Gene Name
spectrin repeat containing nuclear envelope family member 3
Gene Aliases
C14orf49, NET53, Nesp3
UniGene
Hs.354645
Gene Chromosome Location
Chr.14: 95416083-95516693 on Build GRCh38
Transcripts
NM_152592.3 XM_005267377.3 XM_006720063.3 XM_011536515.2 XM_017021056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5713250 c.2385_2387delTCT p.L796delL Chr.14: 95436971-95436973 on Build GRCh38
5713252 c.2385_2387delTCT p.L796delL Chr.14: 95436971-95436973 on Build GRCh38
3386765 c.2528C>G p.S843W Chr.14: 95436830-95436830 on Build GRCh38
4878395 c.2449T>C p.L817L Chr.14: 95436909-95436909 on Build GRCh38
214548 c.2387_2389delTAC p.L796delL Chr.14: 95436969-95436971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746755589 Chr.14: 95437020-95437020 on Build GRCh38
rs751938689 Chr.14: 95436977-95436977 on Build GRCh38
rs757497360 Chr.14: 95436873-95436873 on Build GRCh38
rs763419751 Chr.14: 95436961-95436961 on Build GRCh38
rs778952658 Chr.14: 95436890-95436890 on Build GRCh38
rs776027792 Chr.14: 95436953-95436953 on Build GRCh38
rs555598757 Chr.14: 95436952-95436952 on Build GRCh38
rs764421402 Chr.14: 95436844-95436844 on Build GRCh38
rs747975370 Chr.14: 95436896-95436896 on Build GRCh38
rs748132626 Chr.14: 95437007-95437007 on Build GRCh38
rs759010266 Chr.14: 95436930-95436930 on Build GRCh38
rs768731428 Chr.14: 95437022-95437023 on Build GRCh38
rs200011095 Chr.14: 95436874-95436874 on Build GRCh38
rs758715860 Chr.14: 95436830-95436830 on Build GRCh38
rs777967910 Chr.14: 95436895-95436896 on Build GRCh38
rs76499929 Chr.14: 95436971-95436973 on Build GRCh38
rs369215627 Chr.14: 95436865-95436865 on Build GRCh38
rs971398111 Chr.14: 95436811-95436811 on Build GRCh38
rs764511039 Chr.14: 95436965-95436965 on Build GRCh38
rs769147719 Chr.14: 95437028-95437028 on Build GRCh38
rs772039220 Chr.14: 95436901-95436901 on Build GRCh38
rs200878334 Chr.14: 95437008-95437008 on Build GRCh38
rs751153719 Chr.14: 95436956-95436956 on Build GRCh38
rs763509790 Chr.14: 95437027-95437027 on Build GRCh38
rs974356373 Chr.14: 95436954-95436954 on Build GRCh38
rs376157244 Chr.14: 95436815-95436815 on Build GRCh38
rs375798321 Chr.14: 95437025-95437025 on Build GRCh38
rs183905400 Chr.14: 95436997-95436997 on Build GRCh38
rs745499387 Chr.14: 95437024-95437024 on Build GRCh38
rs762220103 Chr.14: 95437031-95437031 on Build GRCh38
rs202043284 Chr.14: 95436998-95436998 on Build GRCh38
rs200146914 Chr.14: 95437023-95437023 on Build GRCh38
rs749569675 Chr.14: 95436957-95436959 on Build GRCh38
rs757433429 Chr.14: 95436984-95436984 on Build GRCh38
rs765669538 Chr.14: 95436817-95436817 on Build GRCh38
rs531873700 Chr.14: 95436866-95436866 on Build GRCh38
rs540053794 Chr.14: 95436829-95436829 on Build GRCh38
rs776012276 Chr.14: 95436920-95436920 on Build GRCh38
rs201240192 Chr.14: 95436969-95436971 on Build GRCh38
rs773024495 Chr.14: 95436903-95436903 on Build GRCh38
rs759619847 Chr.14: 95437018-95437020 on Build GRCh38
rs770563232 Chr.14: 95436917-95436917 on Build GRCh38
rs909409683 Chr.14: 95436916-95436916 on Build GRCh38
rs57883787 Chr.14: 95437006-95437006 on Build GRCh38
rs537347555 Chr.14: 95436910-95436910 on Build GRCh38
rs754969110 Chr.14: 95436876-95436876 on Build GRCh38
rs370181203 Chr.14: 95437026-95437026 on Build GRCh38

View Full Product Details