Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68015273-68015540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH3B1
Entrez Gene ID
221
Gene Name
aldehyde dehydrogenase 3 family member B1
Gene Aliases
ALDH4, ALDH7
UniGene
Hs.523841
Gene Chromosome Location
Chr.11: 68008547-68029282 on Build GRCh38
Transcripts
NM_000694.3 NM_001030010.2 NM_001161473.2 NM_001290058.1 NM_001290059.1

SNP Detail

SNP ID Chromosome Location
rs778439890 Chr.11: 68015470-68015470 on Build GRCh38
rs752785703 Chr.11: 68015323-68015323 on Build GRCh38
rs773705334 Chr.11: 68015373-68015373 on Build GRCh38
rs565499278 Chr.11: 68015508-68015508 on Build GRCh38
rs746359070 Chr.11: 68015346-68015346 on Build GRCh38
rs751810727 Chr.11: 68015401-68015401 on Build GRCh38
rs199730372 Chr.11: 68015400-68015400 on Build GRCh38
rs2286170 Chr.11: 68015477-68015477 on Build GRCh38
rs968736130 Chr.11: 68015332-68015332 on Build GRCh38
rs758669358 Chr.11: 68015326-68015326 on Build GRCh38
rs944324536 Chr.11: 68015389-68015389 on Build GRCh38
rs924552858 Chr.11: 68015352-68015352 on Build GRCh38
rs535517326 Chr.11: 68015478-68015478 on Build GRCh38
rs945161750 Chr.11: 68015398-68015398 on Build GRCh38
rs181292499 Chr.11: 68015432-68015432 on Build GRCh38
rs750814122 Chr.11: 68015436-68015436 on Build GRCh38
rs865940739 Chr.11: 68015451-68015451 on Build GRCh38
rs373983642 Chr.11: 68015318-68015318 on Build GRCh38
rs992667162 Chr.11: 68015356-68015356 on Build GRCh38
rs761474661 Chr.11: 68015370-68015370 on Build GRCh38
rs780361353 Chr.11: 68015464-68015464 on Build GRCh38
rs7116695 Chr.11: 68015384-68015384 on Build GRCh38
rs928991817 Chr.11: 68015374-68015374 on Build GRCh38
rs373185377 Chr.11: 68015383-68015383 on Build GRCh38
rs772919973 Chr.11: 68015375-68015375 on Build GRCh38
rs750322243 Chr.11: 68015312-68015312 on Build GRCh38
rs570890777 Chr.11: 68015296-68015296 on Build GRCh38
rs778036759 Chr.11: 68015353-68015353 on Build GRCh38
rs757648022 Chr.11: 68015339-68015339 on Build GRCh38
rs977308588 Chr.11: 68015337-68015337 on Build GRCh38
rs747267919 Chr.11: 68015328-68015328 on Build GRCh38
rs147445707 Chr.11: 68015322-68015322 on Build GRCh38
rs749823581 Chr.11: 68015465-68015465 on Build GRCh38
rs186487054 Chr.11: 68015481-68015481 on Build GRCh38
rs975701184 Chr.11: 68015367-68015367 on Build GRCh38
rs757594635 Chr.11: 68015409-68015409 on Build GRCh38
rs2286171 Chr.11: 68015387-68015387 on Build GRCh38
rs201119062 Chr.11: 68015325-68015325 on Build GRCh38
rs533370288 Chr.11: 68015358-68015358 on Build GRCh38
rs768102067 Chr.11: 68015469-68015469 on Build GRCh38
rs756322019 Chr.11: 68015500-68015500 on Build GRCh38
rs308293 Chr.11: 68015357-68015357 on Build GRCh38
rs781459880 Chr.11: 68015345-68015345 on Build GRCh38
rs766284280 Chr.11: 68015385-68015385 on Build GRCh38
rs757197449 Chr.11: 68015359-68015359 on Build GRCh38
rs528494631 Chr.11: 68015504-68015504 on Build GRCh38
rs771552983 Chr.11: 68015474-68015474 on Build GRCh38
rs369125948 Chr.11: 68015471-68015471 on Build GRCh38
rs770274507 Chr.11: 68015347-68015347 on Build GRCh38
rs760656298 Chr.11: 68015380-68015380 on Build GRCh38
rs868712038 Chr.11: 68015435-68015435 on Build GRCh38

View Full Product Details