Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42480548-42480819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0A1
Entrez Gene ID
535
Gene Name
ATPase H+ transporting V0 subunit a1
Gene Aliases
ATP6N1, ATP6N1A, Stv1, VPP1, Vph1, a1
UniGene
Hs.463074
Gene Chromosome Location
Chr.17: 42458844-42522579 on Build GRCh38
Transcripts
NM_001130020.1 NM_001130021.1 NM_005177.3 XM_005257459.1 XM_005257461.1 XM_005257463.2 XM_011524908.1 XM_017024766.1 XM_017024767.1 XM_017024768.1 XM_017024769.1 XM_017024770.1 XM_017024771.1 XM_017024772.1 XM_017024773.1 XM_017024774.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6586639 c.694_716+3del26 p.? Chr.17: 42480727-42480752 on Build GRCh38
6608027 c.636C>T p.G212G Chr.17: 42480669-42480669 on Build GRCh38
6586638 c.715_737+3del26 p.? Chr.17: 42480727-42480752 on Build GRCh38
6608026 c.657C>T p.G219G Chr.17: 42480669-42480669 on Build GRCh38
3188788 c.642C>T p.Y214Y Chr.17: 42480675-42480675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753688999 Chr.17: 42480694-42480694 on Build GRCh38
rs749612108 Chr.17: 42480634-42480634 on Build GRCh38
rs748693129 Chr.17: 42480695-42480695 on Build GRCh38
rs765961383 Chr.17: 42480669-42480669 on Build GRCh38
rs372334798 Chr.17: 42480783-42480783 on Build GRCh38
rs750456435 Chr.17: 42480738-42480738 on Build GRCh38
rs772861456 Chr.17: 42480793-42480793 on Build GRCh38
rs756631876 Chr.17: 42480764-42480764 on Build GRCh38
rs779113159 Chr.17: 42480778-42480778 on Build GRCh38
rs999446258 Chr.17: 42480671-42480671 on Build GRCh38
rs528394339 Chr.17: 42480777-42480777 on Build GRCh38
rs546073271 Chr.17: 42480723-42480723 on Build GRCh38
rs774107419 Chr.17: 42480661-42480661 on Build GRCh38
rs1043825488 Chr.17: 42480599-42480599 on Build GRCh38
rs753691113 Chr.17: 42480702-42480702 on Build GRCh38
rs145824527 Chr.17: 42480673-42480673 on Build GRCh38
rs768894532 Chr.17: 42480658-42480658 on Build GRCh38
rs199699718 Chr.17: 42480727-42480727 on Build GRCh38
rs953936357 Chr.17: 42480787-42480787 on Build GRCh38
rs569974016 Chr.17: 42480611-42480611 on Build GRCh38
rs766360581 Chr.17: 42480675-42480675 on Build GRCh38
rs556721303 Chr.17: 42480607-42480607 on Build GRCh38
rs368543757 Chr.17: 42480732-42480732 on Build GRCh38
rs780599804 Chr.17: 42480773-42480773 on Build GRCh38
rs998134388 Chr.17: 42480769-42480769 on Build GRCh38
rs902615218 Chr.17: 42480625-42480625 on Build GRCh38
rs41283429 Chr.17: 42480711-42480711 on Build GRCh38
rs764929128 Chr.17: 42480712-42480712 on Build GRCh38
rs946788902 Chr.17: 42480596-42480596 on Build GRCh38
rs371663080 Chr.17: 42480786-42480786 on Build GRCh38
rs779373237 Chr.17: 42480577-42480577 on Build GRCh38
rs772806504 Chr.17: 42480674-42480674 on Build GRCh38

View Full Product Details