Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133263043-133263316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM8
Entrez Gene ID
101
Gene Name
ADAM metallopeptidase domain 8
Gene Aliases
CD156, CD156a, MS2
UniGene
Hs.501574
Gene Chromosome Location
Chr.10: 133262416-133276903 on Build GRCh38
Transcripts
NM_001109.4 NM_001164489.1 NM_001164490.1 XM_011539117.2 XM_017015465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7129292 c.2456G>T p.G819V Chr.10: 133263175-133263175 on Build GRCh38
7315470 c.2431C>T p.P811S Chr.10: 133263200-133263200 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759035466 Chr.10: 133263101-133263101 on Build GRCh38
rs34549109 Chr.10: 133263069-133263070 on Build GRCh38
rs776766935 Chr.10: 133263151-133263151 on Build GRCh38
rs775251845 Chr.10: 133263107-133263107 on Build GRCh38
rs779149952 Chr.10: 133263154-133263154 on Build GRCh38
rs150345190 Chr.10: 133263271-133263271 on Build GRCh38
rs753825484 Chr.10: 133263193-133263193 on Build GRCh38
rs576947417 Chr.10: 133263137-133263137 on Build GRCh38
rs761658180 Chr.10: 133263124-133263124 on Build GRCh38
rs886977109 Chr.10: 133263273-133263273 on Build GRCh38
rs758626065 Chr.10: 133263268-133263268 on Build GRCh38
rs34253049 Chr.10: 133263080-133263080 on Build GRCh38
rs767434175 Chr.10: 133263129-133263129 on Build GRCh38
rs779061298 Chr.10: 133263071-133263071 on Build GRCh38
rs768353887 Chr.10: 133263216-133263216 on Build GRCh38
rs773173731 Chr.10: 133263244-133263244 on Build GRCh38
rs3008318 Chr.10: 133263254-133263254 on Build GRCh38
rs1029935213 Chr.10: 133263085-133263085 on Build GRCh38
rs759662255 Chr.10: 133263245-133263245 on Build GRCh38
rs759335509 Chr.10: 133263250-133263251 on Build GRCh38
rs764347913 Chr.10: 133263187-133263187 on Build GRCh38
rs778000373 Chr.10: 133263258-133263258 on Build GRCh38
rs760526884 Chr.10: 133263190-133263190 on Build GRCh38
rs762887142 Chr.10: 133263109-133263109 on Build GRCh38
rs79659580 Chr.10: 133263177-133263177 on Build GRCh38
rs754395281 Chr.10: 133263197-133263197 on Build GRCh38
rs982838006 Chr.10: 133263088-133263088 on Build GRCh38
rs751898958 Chr.10: 133263280-133263280 on Build GRCh38
rs769600887 Chr.10: 133263105-133263105 on Build GRCh38
rs757610120 Chr.10: 133263285-133263285 on Build GRCh38
rs771053269 Chr.10: 133263150-133263151 on Build GRCh38
rs545726760 Chr.10: 133263143-133263143 on Build GRCh38
rs781693749 Chr.10: 133263174-133263174 on Build GRCh38
rs755015233 Chr.10: 133263151-133263151 on Build GRCh38
rs750205020 Chr.10: 133263188-133263188 on Build GRCh38
rs950372424 Chr.10: 133263144-133263144 on Build GRCh38
rs2230576 Chr.10: 133263092-133263092 on Build GRCh38
rs766234759 Chr.10: 133263191-133263191 on Build GRCh38
rs924107821 Chr.10: 133263120-133263120 on Build GRCh38
rs748546654 Chr.10: 133263075-133263075 on Build GRCh38
rs1016327071 Chr.10: 133263284-133263284 on Build GRCh38
rs556384229 Chr.10: 133263163-133263163 on Build GRCh38
rs778301208 Chr.10: 133263086-133263086 on Build GRCh38
rs397733581 Chr.10: 133263176-133263176 on Build GRCh38
rs998489562 Chr.10: 133263286-133263286 on Build GRCh38
rs745408689 Chr.10: 133263176-133263176 on Build GRCh38
rs906421461 Chr.10: 133263247-133263247 on Build GRCh38
rs200356896 Chr.10: 133263172-133263172 on Build GRCh38
rs554340450 Chr.10: 133263100-133263100 on Build GRCh38
rs755172975 Chr.10: 133263070-133263070 on Build GRCh38
rs775336305 Chr.10: 133263185-133263185 on Build GRCh38
rs767389585 Chr.10: 133263241-133263241 on Build GRCh38
rs778457604 Chr.10: 133263240-133263240 on Build GRCh38
rs763963955 Chr.10: 133263116-133263116 on Build GRCh38
rs561363568 Chr.10: 133263173-133263173 on Build GRCh38
rs770542538 Chr.10: 133263093-133263093 on Build GRCh38
rs58683011 Chr.10: 133263176-133263177 on Build GRCh38
rs546715431 Chr.10: 133263117-133263117 on Build GRCh38
rs576260903 Chr.10: 133263170-133263170 on Build GRCh38
rs774027930 Chr.10: 133263224-133263224 on Build GRCh38

View Full Product Details