Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45692781-45693052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN10
Entrez Gene ID
25814
Gene Name
ataxin 10
Gene Aliases
E46L, HUMEEP, SCA10
UniGene
Hs.475125
Gene Chromosome Location
Chr.22: 45671798-45845307 on Build GRCh38
Transcripts
NM_001167621.1 NM_013236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5393839 c.355C>T p.R119C Chr.22: 45693042-45693042 on Build GRCh38
4610681 c.356G>A p.R119H Chr.22: 45693043-45693043 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1032742759 Chr.22: 45692863-45692863 on Build GRCh38
rs371477412 Chr.22: 45692993-45692993 on Build GRCh38
rs781002957 Chr.22: 45693007-45693007 on Build GRCh38
rs773558014 Chr.22: 45693013-45693013 on Build GRCh38
rs765083131 Chr.22: 45692950-45692950 on Build GRCh38
rs1030016963 Chr.22: 45692938-45692938 on Build GRCh38
rs113957803 Chr.22: 45692858-45692858 on Build GRCh38
rs200591585 Chr.22: 45693014-45693014 on Build GRCh38
rs754610439 Chr.22: 45692994-45692994 on Build GRCh38
rs573643659 Chr.22: 45692892-45692892 on Build GRCh38
rs957019081 Chr.22: 45692919-45692919 on Build GRCh38
rs572331678 Chr.22: 45692805-45692805 on Build GRCh38
rs750416231 Chr.22: 45692960-45692960 on Build GRCh38
rs537225177 Chr.22: 45692812-45692812 on Build GRCh38
rs552503406 Chr.22: 45692984-45692984 on Build GRCh38
rs148464355 Chr.22: 45693025-45693025 on Build GRCh38
rs894451333 Chr.22: 45692814-45692814 on Build GRCh38
rs768439641 Chr.22: 45692991-45692991 on Build GRCh38
rs952983559 Chr.22: 45692942-45692942 on Build GRCh38
rs771377877 Chr.22: 45693016-45693016 on Build GRCh38
rs1023882198 Chr.22: 45692952-45692952 on Build GRCh38
rs879559723 Chr.22: 45692922-45692922 on Build GRCh38
rs956469668 Chr.22: 45692868-45692870 on Build GRCh38
rs61733598 Chr.22: 45693008-45693008 on Build GRCh38
rs998535740 Chr.22: 45692845-45692845 on Build GRCh38
rs770258259 Chr.22: 45693009-45693009 on Build GRCh38
rs757135643 Chr.22: 45692948-45692948 on Build GRCh38
rs1031843869 Chr.22: 45692866-45692866 on Build GRCh38
rs535889807 Chr.22: 45692947-45692947 on Build GRCh38
rs749614004 Chr.22: 45692978-45692978 on Build GRCh38
rs185007268 Chr.22: 45692806-45692806 on Build GRCh38
rs975818721 Chr.22: 45692909-45692909 on Build GRCh38
rs757824537 Chr.22: 45692963-45692963 on Build GRCh38
rs867246164 Chr.22: 45692811-45692811 on Build GRCh38
rs573976645 Chr.22: 45692926-45692926 on Build GRCh38
rs41279799 Chr.22: 45692819-45692819 on Build GRCh38

View Full Product Details