Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81665020-81665273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OXLD1
Entrez Gene ID
339229
Gene Name
oxidoreductase like domain containing 1
Gene Aliases
C17orf90
UniGene
Hs.405942
Gene Chromosome Location
Chr.17: 81665036-81666637 on Build GRCh38
Transcripts
NM_001039842.2 NM_001304994.1 NM_001304995.1 NM_001304999.1 NR_130936.1 NR_130937.1 NR_130938.1 NR_130939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
708648 c.436G>A p.G146R Chr.17: 81665209-81665209 on Build GRCh38
4846201 c.437G>A p.G146E Chr.17: 81665208-81665208 on Build GRCh38
6736332 c.374A>G p.H125R Chr.17: 81665271-81665271 on Build GRCh38
3796215 c.382G>C p.D128H Chr.17: 81665263-81665263 on Build GRCh38
6356287 c.435C>T p.C145C Chr.17: 81665210-81665210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528133431 Chr.17: 81665052-81665052 on Build GRCh38
rs947970588 Chr.17: 81665069-81665070 on Build GRCh38
rs370129678 Chr.17: 81665209-81665209 on Build GRCh38
rs779309581 Chr.17: 81665242-81665242 on Build GRCh38
rs781438420 Chr.17: 81665226-81665226 on Build GRCh38
rs1032811016 Chr.17: 81665167-81665167 on Build GRCh38
rs757472919 Chr.17: 81665172-81665172 on Build GRCh38
rs888524357 Chr.17: 81665147-81665147 on Build GRCh38
rs1046755720 Chr.17: 81665077-81665077 on Build GRCh38
rs571134572 Chr.17: 81665152-81665152 on Build GRCh38
rs971796778 Chr.17: 81665223-81665223 on Build GRCh38
rs770566999 Chr.17: 81665196-81665196 on Build GRCh38
rs755487099 Chr.17: 81665231-81665231 on Build GRCh38
rs900814495 Chr.17: 81665135-81665135 on Build GRCh38
rs546696618 Chr.17: 81665106-81665106 on Build GRCh38
rs770349748 Chr.17: 81665249-81665249 on Build GRCh38
rs377740086 Chr.17: 81665253-81665253 on Build GRCh38
rs906475869 Chr.17: 81665126-81665126 on Build GRCh38
rs759257512 Chr.17: 81665201-81665201 on Build GRCh38
rs866000245 Chr.17: 81665246-81665246 on Build GRCh38
rs573397609 Chr.17: 81665208-81665208 on Build GRCh38
rs371953459 Chr.17: 81665177-81665177 on Build GRCh38
rs560433620 Chr.17: 81665178-81665178 on Build GRCh38
rs776294291 Chr.17: 81665200-81665200 on Build GRCh38
rs373706185 Chr.17: 81665160-81665160 on Build GRCh38
rs569616173 Chr.17: 81665176-81665176 on Build GRCh38
rs202131274 Chr.17: 81665161-81665161 on Build GRCh38
rs769109407 Chr.17: 81665180-81665180 on Build GRCh38
rs745501777 Chr.17: 81665255-81665255 on Build GRCh38
rs751661019 Chr.17: 81665165-81665165 on Build GRCh38
rs780553027 Chr.17: 81665228-81665228 on Build GRCh38
rs200385200 Chr.17: 81665247-81665247 on Build GRCh38
rs756489668 Chr.17: 81665227-81665227 on Build GRCh38
rs760150517 Chr.17: 81665190-81665190 on Build GRCh38
rs763982616 Chr.17: 81665211-81665211 on Build GRCh38
rs776204597 Chr.17: 81665254-81665254 on Build GRCh38
rs772747402 Chr.17: 81665182-81665182 on Build GRCh38
rs781678635 Chr.17: 81665134-81665134 on Build GRCh38
rs371162588 Chr.17: 81665158-81665158 on Build GRCh38
rs779355088 Chr.17: 81665186-81665187 on Build GRCh38
rs137869680 Chr.17: 81665217-81665217 on Build GRCh38

View Full Product Details