Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35726393-35726666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOL5
Entrez Gene ID
80831
Gene Name
apolipoprotein L5
Gene Aliases
APOL-V, APOLV
UniGene
Hs.326561
Gene Chromosome Location
Chr.22: 35708424-35730279 on Build GRCh38
Transcripts
NM_030642.1 XM_006724321.3 XM_017028945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4103784 c.469G>A p.A157T Chr.22: 35726537-35726537 on Build GRCh38
7287922 c.478C>T p.P160S Chr.22: 35726546-35726546 on Build GRCh38
4996073 c.527T>C p.L176S Chr.22: 35726595-35726595 on Build GRCh38
6602456 c.340G>T p.E114* Chr.22: 35726408-35726408 on Build GRCh38
278922 c.371C>T p.A124V Chr.22: 35726439-35726439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761542526 Chr.22: 35726468-35726468 on Build GRCh38
rs1053008911 Chr.22: 35726510-35726510 on Build GRCh38
rs561182898 Chr.22: 35726465-35726465 on Build GRCh38
rs755605089 Chr.22: 35726578-35726578 on Build GRCh38
rs199758454 Chr.22: 35726490-35726490 on Build GRCh38
rs981750016 Chr.22: 35726508-35726508 on Build GRCh38
rs1035014551 Chr.22: 35726602-35726602 on Build GRCh38
rs142340026 Chr.22: 35726461-35726461 on Build GRCh38
rs772600028 Chr.22: 35726483-35726483 on Build GRCh38
rs749957392 Chr.22: 35726460-35726460 on Build GRCh38
rs767117995 Chr.22: 35726565-35726565 on Build GRCh38
rs572539048 Chr.22: 35726451-35726451 on Build GRCh38
rs775428622 Chr.22: 35726507-35726507 on Build GRCh38
rs763687163 Chr.22: 35726440-35726440 on Build GRCh38
rs141273692 Chr.22: 35726501-35726501 on Build GRCh38
rs758778060 Chr.22: 35726633-35726633 on Build GRCh38
rs749929863 Chr.22: 35726575-35726575 on Build GRCh38
rs773932855 Chr.22: 35726551-35726551 on Build GRCh38
rs762692848 Chr.22: 35726517-35726517 on Build GRCh38
rs776766391 Chr.22: 35726500-35726500 on Build GRCh38
rs17723764 Chr.22: 35726470-35726470 on Build GRCh38
rs376424899 Chr.22: 35726494-35726494 on Build GRCh38
rs761439903 Chr.22: 35726562-35726562 on Build GRCh38
rs752504367 Chr.22: 35726439-35726439 on Build GRCh38
rs148268031 Chr.22: 35726484-35726484 on Build GRCh38
rs1013734604 Chr.22: 35726591-35726591 on Build GRCh38
rs887484530 Chr.22: 35726462-35726462 on Build GRCh38
rs551718865 Chr.22: 35726613-35726613 on Build GRCh38
rs73411940 Chr.22: 35726454-35726454 on Build GRCh38
rs558909936 Chr.22: 35726639-35726639 on Build GRCh38
rs960689578 Chr.22: 35726621-35726621 on Build GRCh38
rs532000835 Chr.22: 35726529-35726529 on Build GRCh38
rs763877157 Chr.22: 35726518-35726518 on Build GRCh38
rs769747567 Chr.22: 35726505-35726505 on Build GRCh38
rs765152614 Chr.22: 35726434-35726434 on Build GRCh38
rs1044243474 Chr.22: 35726421-35726421 on Build GRCh38
rs572416546 Chr.22: 35726433-35726433 on Build GRCh38
rs753261485 Chr.22: 35726622-35726622 on Build GRCh38
rs776375825 Chr.22: 35726423-35726423 on Build GRCh38
rs1046110886 Chr.22: 35726628-35726628 on Build GRCh38
rs751240367 Chr.22: 35726448-35726448 on Build GRCh38

View Full Product Details