Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 91220868-91221097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUH
Entrez Gene ID
549
Gene Name
AU RNA binding protein/enoyl-CoA hydratase
Gene Aliases
-
UniGene
Hs.175905
Gene Chromosome Location
Chr.9: 91213815-91361969 on Build GRCh38
Transcripts
NM_001306190.1 NM_001698.2 XM_005252066.3 XM_005252067.4 XM_005252069.4 XM_006717150.3 XM_011518802.2 XM_017014849.1 XM_017014850.1 XM_017014851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3221739 c.721C>T p.R241* Chr.9: 91220927-91220927 on Build GRCh38
1111005 c.727C>A p.L243I Chr.9: 91220921-91220921 on Build GRCh38
3908807 c.712T>C p.F238L Chr.9: 91220936-91220936 on Build GRCh38
1111007 c.680C>T p.A227V Chr.9: 91220968-91220968 on Build GRCh38
3908805 c.730G>A p.D244N Chr.9: 91220918-91220918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141700491 Chr.9: 91220969-91220969 on Build GRCh38
rs539768744 Chr.9: 91221065-91221065 on Build GRCh38
rs777963106 Chr.9: 91220988-91220988 on Build GRCh38
rs761519675 Chr.9: 91220955-91220955 on Build GRCh38
rs148157596 Chr.9: 91220972-91220972 on Build GRCh38
rs773055968 Chr.9: 91220968-91220968 on Build GRCh38
rs141110052 Chr.9: 91220991-91220991 on Build GRCh38
rs781142774 Chr.9: 91220988-91220988 on Build GRCh38
rs200472338 Chr.9: 91220997-91220997 on Build GRCh38
rs200212229 Chr.9: 91220927-91220927 on Build GRCh38
rs746917200 Chr.9: 91221017-91221017 on Build GRCh38
rs147928826 Chr.9: 91220939-91220939 on Build GRCh38
rs146636009 Chr.9: 91220919-91220919 on Build GRCh38
rs780964098 Chr.9: 91220994-91220994 on Build GRCh38
rs547093326 Chr.9: 91220910-91220910 on Build GRCh38
rs769329560 Chr.9: 91221005-91221006 on Build GRCh38
rs769894315 Chr.9: 91220929-91220929 on Build GRCh38
rs73497143 Chr.9: 91220986-91220986 on Build GRCh38
rs779230138 Chr.9: 91220980-91220980 on Build GRCh38
rs143768042 Chr.9: 91220970-91220970 on Build GRCh38
rs768508720 Chr.9: 91221031-91221031 on Build GRCh38
rs902491412 Chr.9: 91221030-91221030 on Build GRCh38
rs199879150 Chr.9: 91221000-91221000 on Build GRCh38
rs150082246 Chr.9: 91220993-91220993 on Build GRCh38
rs752072326 Chr.9: 91220902-91220902 on Build GRCh38
rs752044125 Chr.9: 91220971-91220971 on Build GRCh38
rs757748207 Chr.9: 91220993-91220994 on Build GRCh38
rs753032086 Chr.9: 91220981-91220981 on Build GRCh38
rs755299132 Chr.9: 91220906-91220906 on Build GRCh38
rs997680367 Chr.9: 91220903-91220903 on Build GRCh38
rs747402902 Chr.9: 91220894-91220895 on Build GRCh38
rs748551955 Chr.9: 91221061-91221061 on Build GRCh38
rs202182817 Chr.9: 91220918-91220918 on Build GRCh38
rs780040124 Chr.9: 91220928-91220928 on Build GRCh38
rs773262900 Chr.9: 91220936-91220936 on Build GRCh38
rs745577824 Chr.9: 91221005-91221008 on Build GRCh38
rs768440462 Chr.9: 91220932-91220932 on Build GRCh38
rs769547921 Chr.9: 91220959-91220959 on Build GRCh38

View Full Product Details