Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101655852-101656079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX2
Entrez Gene ID
9823
Gene Name
armadillo repeat containing, X-linked 2
Gene Aliases
ALEX2, GASP9
UniGene
Hs.48924
Gene Chromosome Location
Chr.X: 101655281-101659891 on Build GRCh38
Transcripts
NM_001282231.1 NM_014782.6 NM_177949.3 XM_005278109.1 XM_005278110.1 XM_005278111.1 XM_005278113.1 XM_005278114.1 XM_005278115.1 XM_005278116.1 XM_005278117.1 XM_011531071.1 XM_011531072.1 XM_017029987.1 XM_017029988.1 XM_017029989.1 XM_017029990.1 XM_017029991.1 XM_017029992.1 XM_017029993.1 XM_017029994.1 XM_017029995.1 XM_017029996.1 XM_017029997.1

SNP Detail

SNP ID Chromosome Location
rs782671204 Chr.X: 101655896-101655896 on Build GRCh38
rs781968607 Chr.X: 101655920-101655920 on Build GRCh38
rs781929040 Chr.X: 101655957-101655957 on Build GRCh38
rs782182924 Chr.X: 101655885-101655885 on Build GRCh38
rs73568864 Chr.X: 101655930-101655930 on Build GRCh38
rs782382207 Chr.X: 101655915-101655915 on Build GRCh38
rs199503721 Chr.X: 101656019-101656019 on Build GRCh38
rs782335187 Chr.X: 101655942-101655942 on Build GRCh38
rs782089774 Chr.X: 101656036-101656036 on Build GRCh38
rs192734301 Chr.X: 101656045-101656045 on Build GRCh38
rs782171989 Chr.X: 101655984-101655984 on Build GRCh38
rs782261396 Chr.X: 101655910-101655910 on Build GRCh38
rs782551420 Chr.X: 101655888-101655888 on Build GRCh38
rs782800621 Chr.X: 101655988-101655988 on Build GRCh38

View Full Product Details