Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51263952-51264219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIGLECL1
Entrez Gene ID
284369
Gene Name
SIGLEC family like 1
Gene Aliases
C19orf75, SIGLEC23P, SIGLECP7
UniGene
Hs.381087
Gene Chromosome Location
Chr.19: 51251064-51269330 on Build GRCh38
Transcripts
NM_001308437.1 NM_173635.2 NR_102324.1 XM_005258799.4 XM_011526821.2 XM_017026647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5028614 c.3G>A p.M1I Chr.19: 51264075-51264075 on Build GRCh38
4755169 c.17A>C p.Q6P Chr.19: 51264089-51264089 on Build GRCh38
6778472 c.22+4A>C p.? Chr.19: 51264098-51264098 on Build GRCh38
7326221 c.22+7A>T p.? Chr.19: 51264101-51264101 on Build GRCh38
475111 c.22G>T p.V8L Chr.19: 51264094-51264094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746854466 Chr.19: 51264050-51264050 on Build GRCh38
rs961814818 Chr.19: 51263976-51263976 on Build GRCh38
rs780212929 Chr.19: 51264026-51264026 on Build GRCh38
rs762693615 Chr.19: 51264088-51264088 on Build GRCh38
rs200972320 Chr.19: 51264143-51264143 on Build GRCh38
rs368235420 Chr.19: 51264157-51264157 on Build GRCh38
rs958073507 Chr.19: 51264164-51264164 on Build GRCh38
rs143635001 Chr.19: 51264074-51264074 on Build GRCh38
rs367828225 Chr.19: 51264132-51264132 on Build GRCh38
rs544119121 Chr.19: 51264006-51264006 on Build GRCh38
rs761207263 Chr.19: 51264104-51264104 on Build GRCh38
rs200316667 Chr.19: 51264103-51264103 on Build GRCh38
rs370870695 Chr.19: 51264112-51264112 on Build GRCh38
rs562329475 Chr.19: 51264055-51264055 on Build GRCh38
rs1028197977 Chr.19: 51264116-51264116 on Build GRCh38
rs901228444 Chr.19: 51264165-51264165 on Build GRCh38
rs368337741 Chr.19: 51264002-51264002 on Build GRCh38
rs761373555 Chr.19: 51264095-51264098 on Build GRCh38
rs764687374 Chr.19: 51264107-51264107 on Build GRCh38
rs374273575 Chr.19: 51264076-51264076 on Build GRCh38
rs747929636 Chr.19: 51264068-51264068 on Build GRCh38
rs376428269 Chr.19: 51264102-51264102 on Build GRCh38
rs748300617 Chr.19: 51264152-51264152 on Build GRCh38
rs758359870 Chr.19: 51264025-51264025 on Build GRCh38
rs1040667529 Chr.19: 51264056-51264056 on Build GRCh38
rs148098048 Chr.19: 51264081-51264081 on Build GRCh38
rs1011000779 Chr.19: 51264173-51264173 on Build GRCh38
rs996812356 Chr.19: 51264166-51264166 on Build GRCh38
rs75883413 Chr.19: 51263975-51263975 on Build GRCh38
rs765656256 Chr.19: 51264122-51264122 on Build GRCh38
rs143751925 Chr.19: 51263995-51263995 on Build GRCh38
rs750571529 Chr.19: 51264137-51264137 on Build GRCh38
rs971794008 Chr.19: 51263984-51263984 on Build GRCh38

View Full Product Details