Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 31448242-31448489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTD2
Entrez Gene ID
112487
Gene Name
D-tyrosyl-tRNA deacylase 2 (putative)
Gene Aliases
C14orf126
UniGene
Hs.116014
Gene Chromosome Location
Chr.14: 31446036-31457474 on Build GRCh38
Transcripts
NM_080664.2 NR_110045.1

Gene Symbol
LOC101927124
Entrez Gene ID
101927124
Gene Name
uncharacterized LOC101927124
Gene Aliases
-
UniGene
Hs.647561
Gene Chromosome Location
Chr.14: 31420758-31452883 on Build GRCh38
Transcripts
NM_080664.2 NR_110045.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6054650 c.305G>T p.R102I Chr.14: 31448331-31448331 on Build GRCh38
1197378 c.232G>A p.V78I Chr.14: 31448404-31448404 on Build GRCh38
955320 c.234C>A p.V78V Chr.14: 31448402-31448402 on Build GRCh38
6649935 c.373delC p.L125fs*6 Chr.14: 31448263-31448263 on Build GRCh38
955319 c.326A>T p.N109I Chr.14: 31448310-31448310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780719164 Chr.14: 31448309-31448309 on Build GRCh38
rs982828891 Chr.14: 31448444-31448444 on Build GRCh38
rs945525653 Chr.14: 31448451-31448451 on Build GRCh38
rs369632474 Chr.14: 31448402-31448402 on Build GRCh38
rs992179046 Chr.14: 31448298-31448298 on Build GRCh38
rs868811639 Chr.14: 31448410-31448410 on Build GRCh38
rs751099524 Chr.14: 31448303-31448303 on Build GRCh38
rs777747350 Chr.14: 31448362-31448362 on Build GRCh38
rs754588881 Chr.14: 31448305-31448305 on Build GRCh38
rs763409265 Chr.14: 31448435-31448435 on Build GRCh38
rs759633953 Chr.14: 31448418-31448419 on Build GRCh38
rs891067537 Chr.14: 31448314-31448315 on Build GRCh38
rs143796407 Chr.14: 31448325-31448325 on Build GRCh38
rs553222866 Chr.14: 31448294-31448294 on Build GRCh38
rs566857257 Chr.14: 31448398-31448398 on Build GRCh38
rs754501876 Chr.14: 31448459-31448459 on Build GRCh38
rs775647096 Chr.14: 31448385-31448385 on Build GRCh38
rs755925923 Chr.14: 31448335-31448335 on Build GRCh38
rs771025566 Chr.14: 31448384-31448384 on Build GRCh38
rs773589527 Chr.14: 31448434-31448434 on Build GRCh38
rs749193818 Chr.14: 31448378-31448378 on Build GRCh38
rs765770820 Chr.14: 31448279-31448279 on Build GRCh38
rs371614838 Chr.14: 31448428-31448428 on Build GRCh38
rs758877503 Chr.14: 31448445-31448445 on Build GRCh38
rs368157537 Chr.14: 31448365-31448365 on Build GRCh38
rs200009799 Chr.14: 31448455-31448455 on Build GRCh38
rs1013148465 Chr.14: 31448377-31448377 on Build GRCh38
rs376668876 Chr.14: 31448301-31448301 on Build GRCh38
rs747100424 Chr.14: 31448393-31448393 on Build GRCh38
rs959233807 Chr.14: 31448293-31448295 on Build GRCh38
rs551899697 Chr.14: 31448296-31448296 on Build GRCh38
rs765568632 Chr.14: 31448432-31448432 on Build GRCh38
rs766875142 Chr.14: 31448439-31448439 on Build GRCh38

View Full Product Details