Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28759394-28759659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM222B
Entrez Gene ID
55731
Gene Name
family with sequence similarity 222 member B
Gene Aliases
C17orf63
UniGene
Hs.564533
Gene Chromosome Location
Chr.17: 28755978-28855036 on Build GRCh38
Transcripts
NM_001077498.2 NM_001288631.1 NM_001288632.1 NM_001288633.1 NM_001288634.1 NM_001288635.1 NM_001288636.1 NM_001288637.1 NM_001288638.1 NM_001288639.1 NM_001288640.1 NM_018182.3 XM_011524986.2 XM_011524987.1 XM_011524988.2 XM_017024831.1 XM_017024832.1 XM_017024833.1 XM_017024834.1 XM_017024835.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416263 c.538C>T p.P180S Chr.17: 28759421-28759421 on Build GRCh38
2801028 c.471C>T p.A157A Chr.17: 28759488-28759488 on Build GRCh38
6436999 c.500C>T p.A167V Chr.17: 28759459-28759459 on Build GRCh38
3819157 c.438C>G p.H146Q Chr.17: 28759521-28759521 on Build GRCh38
2801038 c.369G>C p.L123F Chr.17: 28759590-28759590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547950785 Chr.17: 28759435-28759435 on Build GRCh38
rs200961802 Chr.17: 28759417-28759417 on Build GRCh38
rs566306998 Chr.17: 28759446-28759446 on Build GRCh38
rs778797341 Chr.17: 28759607-28759607 on Build GRCh38
rs749027205 Chr.17: 28759464-28759464 on Build GRCh38
rs367610761 Chr.17: 28759557-28759557 on Build GRCh38
rs150994811 Chr.17: 28759447-28759447 on Build GRCh38
rs375084101 Chr.17: 28759606-28759606 on Build GRCh38
rs774687868 Chr.17: 28759498-28759498 on Build GRCh38
rs765630646 Chr.17: 28759566-28759566 on Build GRCh38
rs753858204 Chr.17: 28759603-28759603 on Build GRCh38
rs748336987 Chr.17: 28759499-28759499 on Build GRCh38
rs771090558 Chr.17: 28759620-28759620 on Build GRCh38
rs760810702 Chr.17: 28759543-28759543 on Build GRCh38
rs11371891 Chr.17: 28759437-28759438 on Build GRCh38
rs755435752 Chr.17: 28759425-28759425 on Build GRCh38
rs776972509 Chr.17: 28759624-28759624 on Build GRCh38
rs532126220 Chr.17: 28759414-28759414 on Build GRCh38
rs397746964 Chr.17: 28759436-28759436 on Build GRCh38
rs184608986 Chr.17: 28759600-28759600 on Build GRCh38
rs767750259 Chr.17: 28759412-28759412 on Build GRCh38
rs192183191 Chr.17: 28759594-28759594 on Build GRCh38
rs933794460 Chr.17: 28759522-28759522 on Build GRCh38
rs369509726 Chr.17: 28759608-28759608 on Build GRCh38
rs772442770 Chr.17: 28759521-28759521 on Build GRCh38
rs770300215 Chr.17: 28759637-28759637 on Build GRCh38
rs529852734 Chr.17: 28759423-28759423 on Build GRCh38
rs371932859 Chr.17: 28759575-28759575 on Build GRCh38
rs763721994 Chr.17: 28759430-28759430 on Build GRCh38
rs369677017 Chr.17: 28759569-28759569 on Build GRCh38
rs755940006 Chr.17: 28759460-28759460 on Build GRCh38
rs764154769 Chr.17: 28759454-28759454 on Build GRCh38
rs538977003 Chr.17: 28759633-28759633 on Build GRCh38
rs755710267 Chr.17: 28759598-28759598 on Build GRCh38
rs772351853 Chr.17: 28759611-28759611 on Build GRCh38
rs375394898 Chr.17: 28759565-28759565 on Build GRCh38
rs775766158 Chr.17: 28759638-28759638 on Build GRCh38
rs930697931 Chr.17: 28759514-28759514 on Build GRCh38
rs996332099 Chr.17: 28759581-28759581 on Build GRCh38
rs780108619 Chr.17: 28759461-28759461 on Build GRCh38
rs749898291 Chr.17: 28759595-28759595 on Build GRCh38
rs773105560 Chr.17: 28759540-28759540 on Build GRCh38
rs397723149 Chr.17: 28759435-28759435 on Build GRCh38
rs768625209 Chr.17: 28759474-28759474 on Build GRCh38
rs771335348 Chr.17: 28759552-28759552 on Build GRCh38
rs755315366 Chr.17: 28759463-28759498 on Build GRCh38
rs200113013 Chr.17: 28759422-28759422 on Build GRCh38
rs761035119 Chr.17: 28759421-28759421 on Build GRCh38
rs747044894 Chr.17: 28759614-28759614 on Build GRCh38
rs397724969 Chr.17: 28759437-28759437 on Build GRCh38

View Full Product Details