Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6590792-6591027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD70
Entrez Gene ID
970
Gene Name
CD70 molecule
Gene Aliases
CD27L, CD27LG, TNFSF7, TNLG8A
UniGene
Hs.501497
Gene Chromosome Location
Chr.19: 6581646-6591152 on Build GRCh38
Transcripts
NM_001252.4 XM_017027523.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4963662 c.70G>T p.V24F Chr.19: 6590933-6590933 on Build GRCh38
6623882 c.162G>A p.G54G Chr.19: 6590841-6590841 on Build GRCh38
2149206 c.140_141delAG p.Q47fs*12 Chr.19: 6590862-6590863 on Build GRCh38
6152438 c.81C>A p.V27V Chr.19: 6590922-6590922 on Build GRCh38
6623880 c.83C>T p.A28V Chr.19: 6590920-6590920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147810644 Chr.19: 6590940-6590940 on Build GRCh38
rs760044216 Chr.19: 6590896-6590896 on Build GRCh38
rs960367796 Chr.19: 6590865-6590865 on Build GRCh38
rs762476680 Chr.19: 6590980-6590980 on Build GRCh38
rs747119880 Chr.19: 6590934-6590934 on Build GRCh38
rs1034616745 Chr.19: 6590854-6590854 on Build GRCh38
rs370658748 Chr.19: 6590992-6590992 on Build GRCh38
rs145065586 Chr.19: 6590955-6590955 on Build GRCh38
rs765829846 Chr.19: 6590984-6590984 on Build GRCh38
rs144259158 Chr.19: 6590909-6590909 on Build GRCh38
rs199561952 Chr.19: 6590862-6590862 on Build GRCh38
rs758517850 Chr.19: 6590835-6590835 on Build GRCh38
rs776250575 Chr.19: 6590977-6590977 on Build GRCh38
rs750834819 Chr.19: 6590989-6590989 on Build GRCh38
rs904974032 Chr.19: 6590837-6590837 on Build GRCh38
rs750812571 Chr.19: 6590921-6590921 on Build GRCh38
rs984672251 Chr.19: 6590913-6590913 on Build GRCh38
rs201175134 Chr.19: 6590831-6590831 on Build GRCh38
rs770660325 Chr.19: 6590876-6590876 on Build GRCh38
rs750225124 Chr.19: 6590861-6590863 on Build GRCh38
rs1001927429 Chr.19: 6590853-6590853 on Build GRCh38
rs749269246 Chr.19: 6590944-6590944 on Build GRCh38
rs146389258 Chr.19: 6590852-6590852 on Build GRCh38
rs148772362 Chr.19: 6590900-6590900 on Build GRCh38
rs777583938 Chr.19: 6590864-6590864 on Build GRCh38
rs149645346 Chr.19: 6590867-6590867 on Build GRCh38
rs935488818 Chr.19: 6590824-6590824 on Build GRCh38
rs898150658 Chr.19: 6590872-6590872 on Build GRCh38
rs767072737 Chr.19: 6590894-6590894 on Build GRCh38
rs767118220 Chr.19: 6590823-6590823 on Build GRCh38
rs765737694 Chr.19: 6590920-6590920 on Build GRCh38
rs891353796 Chr.19: 6590844-6590844 on Build GRCh38
rs151004469 Chr.19: 6590959-6590959 on Build GRCh38
rs774936872 Chr.19: 6590964-6590964 on Build GRCh38
rs764665507 Chr.19: 6590897-6590897 on Build GRCh38
rs534715855 Chr.19: 6590942-6590942 on Build GRCh38
rs1026268289 Chr.19: 6590888-6590888 on Build GRCh38
rs774106387 Chr.19: 6590882-6590882 on Build GRCh38
rs780084590 Chr.19: 6590930-6590930 on Build GRCh38
rs369190575 Chr.19: 6590960-6590960 on Build GRCh38
rs749109850 Chr.19: 6590875-6590875 on Build GRCh38
rs909952305 Chr.19: 6590918-6590918 on Build GRCh38
rs746699223 Chr.19: 6590997-6590997 on Build GRCh38
rs139425055 Chr.19: 6590857-6590857 on Build GRCh38
rs758708878 Chr.19: 6590926-6590926 on Build GRCh38
rs760261716 Chr.19: 6590968-6590968 on Build GRCh38
rs759222859 Chr.19: 6590883-6590883 on Build GRCh38
rs760406449 Chr.19: 6590962-6590963 on Build GRCh38
rs763748622 Chr.19: 6590976-6590976 on Build GRCh38
rs558131192 Chr.19: 6591004-6591004 on Build GRCh38
rs202182551 Chr.19: 6590841-6590841 on Build GRCh38
rs779302321 Chr.19: 6590827-6590827 on Build GRCh38
rs754265248 Chr.19: 6590815-6590815 on Build GRCh38
rs763163424 Chr.19: 6590990-6590990 on Build GRCh38
rs951965861 Chr.19: 6590901-6590901 on Build GRCh38
rs139447880 Chr.19: 6590951-6590951 on Build GRCh38
rs757627362 Chr.19: 6590819-6590819 on Build GRCh38

View Full Product Details