Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36200205-36200464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRPF3
Entrez Gene ID
27154
Gene Name
bromodomain and PHD finger containing 3
Gene Aliases
-
UniGene
Hs.520096
Gene Chromosome Location
Chr.6: 36196773-36232790 on Build GRCh38
Transcripts
NM_015695.2 XM_005249010.2 XM_005249011.3 XM_011514489.1 XM_011514490.1 XM_011514491.1 XM_011514492.1 XM_017010742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4541967 c.30G>A p.Q10Q Chr.6: 36200352-36200352 on Build GRCh38
3745208 c.99A>G p.T33T Chr.6: 36200421-36200421 on Build GRCh38
5782861 c.56C>G p.S19C Chr.6: 36200378-36200378 on Build GRCh38
4432781 c.26G>A p.R9Q Chr.6: 36200348-36200348 on Build GRCh38
1078478 c.14G>A p.R5H Chr.6: 36200336-36200336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780866046 Chr.6: 36200437-36200437 on Build GRCh38
rs757939709 Chr.6: 36200422-36200422 on Build GRCh38
rs756476074 Chr.6: 36200331-36200331 on Build GRCh38
rs748953638 Chr.6: 36200339-36200339 on Build GRCh38
rs544416359 Chr.6: 36200413-36200413 on Build GRCh38
rs774598096 Chr.6: 36200373-36200373 on Build GRCh38
rs979645029 Chr.6: 36200421-36200421 on Build GRCh38
rs540433573 Chr.6: 36200321-36200321 on Build GRCh38
rs746975010 Chr.6: 36200431-36200431 on Build GRCh38
rs756304125 Chr.6: 36200366-36200366 on Build GRCh38
rs533389243 Chr.6: 36200433-36200433 on Build GRCh38
rs192882745 Chr.6: 36200252-36200252 on Build GRCh38
rs562183508 Chr.6: 36200376-36200376 on Build GRCh38
rs1017696805 Chr.6: 36200246-36200246 on Build GRCh38
rs761948959 Chr.6: 36200375-36200375 on Build GRCh38
rs945255339 Chr.6: 36200391-36200391 on Build GRCh38
rs376814623 Chr.6: 36200336-36200336 on Build GRCh38
rs772019126 Chr.6: 36200358-36200358 on Build GRCh38
rs778397237 Chr.6: 36200348-36200348 on Build GRCh38
rs925072873 Chr.6: 36200424-36200424 on Build GRCh38
rs751624954 Chr.6: 36200312-36200312 on Build GRCh38
rs765523099 Chr.6: 36200283-36200283 on Build GRCh38
rs754162111 Chr.6: 36200387-36200387 on Build GRCh38
rs199876915 Chr.6: 36200381-36200381 on Build GRCh38
rs757856246 Chr.6: 36200313-36200313 on Build GRCh38
rs750835692 Chr.6: 36200320-36200320 on Build GRCh38
rs748044267 Chr.6: 36200271-36200272 on Build GRCh38
rs776470804 Chr.6: 36200368-36200368 on Build GRCh38
rs754637960 Chr.6: 36200401-36200401 on Build GRCh38
rs747598121 Chr.6: 36200351-36200351 on Build GRCh38
rs184803483 Chr.6: 36200295-36200295 on Build GRCh38
rs202166840 Chr.6: 36200359-36200359 on Build GRCh38
rs139389902 Chr.6: 36200355-36200355 on Build GRCh38
rs768092295 Chr.6: 36200319-36200319 on Build GRCh38
rs371915331 Chr.6: 36200365-36200365 on Build GRCh38
rs202108091 Chr.6: 36200423-36200423 on Build GRCh38
rs1031408587 Chr.6: 36200327-36200327 on Build GRCh38
rs764356550 Chr.6: 36200369-36200369 on Build GRCh38
rs1030318549 Chr.6: 36200232-36200232 on Build GRCh38
rs778487176 Chr.6: 36200409-36200409 on Build GRCh38
rs942103247 Chr.6: 36200380-36200380 on Build GRCh38
rs951616929 Chr.6: 36200255-36200255 on Build GRCh38
rs760338400 Chr.6: 36200280-36200280 on Build GRCh38
rs535488863 Chr.6: 36200332-36200332 on Build GRCh38
rs746931056 Chr.6: 36200363-36200363 on Build GRCh38
rs754656237 Chr.6: 36200347-36200347 on Build GRCh38
rs142960302 Chr.6: 36200384-36200384 on Build GRCh38

View Full Product Details