Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26387826-26388083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN2A2
Entrez Gene ID
10385
Gene Name
butyrophilin subfamily 2 member A2
Gene Aliases
BT2.2, BTF2, BTN2.2
UniGene
Hs.373938
Gene Chromosome Location
Chr.6: 26382891-26394874 on Build GRCh38
Transcripts
NM_001197237.1 NM_001197238.1 NM_001197239.1 NM_001197240.1 NM_006995.4 NM_181531.2 XM_005248797.3 XM_005248798.4 XM_006714953.3 XM_006714955.3 XM_011514228.2 XM_011514229.2 XM_011514231.2 XM_017010166.1 XM_017010167.1 XM_017010168.1 XM_017010169.1 XM_011514268.2

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
NM_001197237.1 NM_001197238.1 NM_001197239.1 NM_001197240.1 NM_006995.4 NM_181531.2 XM_005248797.3 XM_005248798.4 XM_006714953.3 XM_006714955.3 XM_011514228.2 XM_011514229.2 XM_011514231.2 XM_017010166.1 XM_017010167.1 XM_017010168.1 XM_017010169.1 XM_011514268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7179327 c.466G>A p.E156K Chr.6: 26388036-26388036 on Build GRCh38
5835161 c.457C>A p.P153T Chr.6: 26388027-26388027 on Build GRCh38
5156925 c.506_516del11 p.C169fs*16 Chr.6: 26388076-26388086 on Build GRCh38
6479960 c.448G>T p.G150W Chr.6: 26388018-26388018 on Build GRCh38
451037 c.457C>A p.P153T Chr.6: 26388027-26388027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565086056 Chr.6: 26387977-26387977 on Build GRCh38
rs192560206 Chr.6: 26387984-26387984 on Build GRCh38
rs756057337 Chr.6: 26387978-26387978 on Build GRCh38
rs373105995 Chr.6: 26388001-26388001 on Build GRCh38
rs866150577 Chr.6: 26387903-26387903 on Build GRCh38
rs748279471 Chr.6: 26388010-26388010 on Build GRCh38
rs779842080 Chr.6: 26387979-26387979 on Build GRCh38
rs763843005 Chr.6: 26387963-26387963 on Build GRCh38
rs190257407 Chr.6: 26387891-26387891 on Build GRCh38
rs914074634 Chr.6: 26387854-26387854 on Build GRCh38
rs773221205 Chr.6: 26388030-26388030 on Build GRCh38
rs1052478631 Chr.6: 26387971-26387971 on Build GRCh38
rs751147516 Chr.6: 26387966-26387966 on Build GRCh38
rs945645797 Chr.6: 26387928-26387928 on Build GRCh38
rs780808399 Chr.6: 26387973-26387973 on Build GRCh38
rs756847197 Chr.6: 26387969-26387969 on Build GRCh38
rs772146273 Chr.6: 26388027-26388027 on Build GRCh38
rs766942506 Chr.6: 26388061-26388061 on Build GRCh38
rs1029274258 Chr.6: 26387882-26387883 on Build GRCh38
rs142227365 Chr.6: 26388005-26388005 on Build GRCh38
rs888037880 Chr.6: 26387912-26387912 on Build GRCh38
rs573542031 Chr.6: 26387880-26387880 on Build GRCh38
rs760735237 Chr.6: 26388042-26388042 on Build GRCh38
rs996561615 Chr.6: 26387878-26387878 on Build GRCh38
rs994888288 Chr.6: 26388048-26388048 on Build GRCh38

View Full Product Details