Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16799615-16799874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGR2
Entrez Gene ID
10551
Gene Name
anterior gradient 2, protein disulphide isomerase family member
Gene Aliases
AG2, GOB-4, HAG-2, HEL-S-116, PDIA17, XAG-2
UniGene
Hs.530009
Gene Chromosome Location
Chr.7: 16791811-16805114 on Build GRCh38
Transcripts
NM_006408.3 XM_005249581.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5526422 c.346G>A p.E116K Chr.7: 16799788-16799788 on Build GRCh38
5640343 c.367C>A p.Q123K Chr.7: 16799767-16799767 on Build GRCh38
1449759 c.281A>G p.N94S Chr.7: 16799793-16799793 on Build GRCh38
1698377 c.286G>A p.E96K Chr.7: 16799788-16799788 on Build GRCh38
6330008 c.354G>T p.Q118H Chr.7: 16799780-16799780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766257117 Chr.7: 16799697-16799697 on Build GRCh38
rs193271402 Chr.7: 16799787-16799787 on Build GRCh38
rs755715177 Chr.7: 16799830-16799831 on Build GRCh38
rs137929187 Chr.7: 16799799-16799799 on Build GRCh38
rs780402357 Chr.7: 16799740-16799740 on Build GRCh38
rs756281126 Chr.7: 16799733-16799733 on Build GRCh38
rs776432855 Chr.7: 16799788-16799788 on Build GRCh38
rs775286089 Chr.7: 16799804-16799804 on Build GRCh38
rs771675727 Chr.7: 16799758-16799758 on Build GRCh38
rs371645198 Chr.7: 16799712-16799712 on Build GRCh38
rs746500663 Chr.7: 16799773-16799773 on Build GRCh38
rs546661571 Chr.7: 16799674-16799674 on Build GRCh38
rs755291829 Chr.7: 16799749-16799749 on Build GRCh38
rs887497089 Chr.7: 16799701-16799701 on Build GRCh38
rs187476985 Chr.7: 16799670-16799670 on Build GRCh38
rs772874557 Chr.7: 16799770-16799770 on Build GRCh38
rs756231760 Chr.7: 16799821-16799821 on Build GRCh38
rs748462919 Chr.7: 16799844-16799844 on Build GRCh38
rs779390697 Chr.7: 16799751-16799751 on Build GRCh38
rs368570959 Chr.7: 16799794-16799794 on Build GRCh38
rs776682877 Chr.7: 16799699-16799699 on Build GRCh38
rs762870671 Chr.7: 16799805-16799805 on Build GRCh38
rs931754960 Chr.7: 16799671-16799671 on Build GRCh38
rs376401016 Chr.7: 16799725-16799725 on Build GRCh38
rs568093378 Chr.7: 16799724-16799724 on Build GRCh38
rs968086921 Chr.7: 16799838-16799838 on Build GRCh38
rs912866850 Chr.7: 16799812-16799812 on Build GRCh38
rs1005682199 Chr.7: 16799706-16799706 on Build GRCh38
rs766545138 Chr.7: 16799824-16799824 on Build GRCh38
rs750487119 Chr.7: 16799726-16799726 on Build GRCh38
rs759224539 Chr.7: 16799700-16799700 on Build GRCh38
rs367829433 Chr.7: 16799705-16799705 on Build GRCh38
rs896874028 Chr.7: 16799646-16799646 on Build GRCh38
rs755447979 Chr.7: 16799837-16799837 on Build GRCh38
rs372209181 Chr.7: 16799806-16799806 on Build GRCh38
rs770518161 Chr.7: 16799786-16799786 on Build GRCh38
rs75651967 Chr.7: 16799727-16799727 on Build GRCh38
rs935258861 Chr.7: 16799649-16799649 on Build GRCh38
rs761847757 Chr.7: 16799657-16799657 on Build GRCh38
rs750722379 Chr.7: 16799817-16799817 on Build GRCh38
rs748518148 Chr.7: 16799755-16799755 on Build GRCh38
rs749842235 Chr.7: 16799747-16799747 on Build GRCh38
rs754285177 Chr.7: 16799825-16799825 on Build GRCh38
rs549949904 Chr.7: 16799643-16799643 on Build GRCh38

View Full Product Details