Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19851269-19851536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf29
Entrez Gene ID
79680
Gene Name
chromosome 22 open reading frame 29
Gene Aliases
BOP
UniGene
Hs.105642
Gene Chromosome Location
Chr.22: 19846138-19854848 on Build GRCh38
Transcripts
NM_024627.5 NM_053004.2

Gene Symbol
GNB1L
Entrez Gene ID
54584
Gene Name
G protein subunit beta 1 like
Gene Aliases
DGCRK3, FKSG1, GY2, WDR14, WDVCF
UniGene
Hs.105642
Gene Chromosome Location
Chr.22: 19788411-19854939 on Build GRCh38
Transcripts
NM_024627.5 NM_053004.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4933288 c.747A>G p.V249V Chr.22: 19851515-19851515 on Build GRCh38
4974739 c.840C>T p.S280S Chr.22: 19851422-19851422 on Build GRCh38
7281918 c.729G>A p.V243V Chr.22: 19851533-19851533 on Build GRCh38
7429537 c.988G>T p.E330* Chr.22: 19851274-19851274 on Build GRCh38
3414199 c.887C>T p.A296V Chr.22: 19851375-19851375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572316704 Chr.22: 19851463-19851463 on Build GRCh38
rs955702207 Chr.22: 19851366-19851366 on Build GRCh38
rs767842472 Chr.22: 19851459-19851459 on Build GRCh38
rs1021754495 Chr.22: 19851381-19851381 on Build GRCh38
rs779045629 Chr.22: 19851509-19851509 on Build GRCh38
rs940655152 Chr.22: 19851450-19851450 on Build GRCh38
rs146626259 Chr.22: 19851371-19851371 on Build GRCh38
rs758703251 Chr.22: 19851385-19851386 on Build GRCh38
rs77448872 Chr.22: 19851368-19851368 on Build GRCh38
rs756609398 Chr.22: 19851465-19851465 on Build GRCh38
rs777985393 Chr.22: 19851391-19851391 on Build GRCh38
rs776672508 Chr.22: 19851336-19851336 on Build GRCh38
rs751181807 Chr.22: 19851451-19851451 on Build GRCh38
rs759536269 Chr.22: 19851415-19851415 on Build GRCh38
rs749589909 Chr.22: 19851396-19851396 on Build GRCh38
rs113321225 Chr.22: 19851360-19851360 on Build GRCh38
rs770819035 Chr.22: 19851411-19851411 on Build GRCh38
rs746813700 Chr.22: 19851404-19851404 on Build GRCh38
rs1050651990 Chr.22: 19851325-19851325 on Build GRCh38
rs529569822 Chr.22: 19851330-19851330 on Build GRCh38
rs143814574 Chr.22: 19851342-19851342 on Build GRCh38
rs760449959 Chr.22: 19851322-19851322 on Build GRCh38
rs749449305 Chr.22: 19851297-19851297 on Build GRCh38
rs900754378 Chr.22: 19851290-19851290 on Build GRCh38
rs780093858 Chr.22: 19851393-19851393 on Build GRCh38
rs368485950 Chr.22: 19851341-19851341 on Build GRCh38
rs774723815 Chr.22: 19851425-19851425 on Build GRCh38
rs148997148 Chr.22: 19851321-19851321 on Build GRCh38
rs779149275 Chr.22: 19851403-19851403 on Build GRCh38
rs996413505 Chr.22: 19851298-19851298 on Build GRCh38
rs78844806 Chr.22: 19851350-19851350 on Build GRCh38
rs775725263 Chr.22: 19851296-19851296 on Build GRCh38
rs372257446 Chr.22: 19851467-19851467 on Build GRCh38
rs369074705 Chr.22: 19851497-19851497 on Build GRCh38
rs763673672 Chr.22: 19851438-19851438 on Build GRCh38
rs372143693 Chr.22: 19851313-19851313 on Build GRCh38
rs762470768 Chr.22: 19851432-19851432 on Build GRCh38
rs769821927 Chr.22: 19851293-19851293 on Build GRCh38
rs907911373 Chr.22: 19851441-19851441 on Build GRCh38
rs752455357 Chr.22: 19851343-19851343 on Build GRCh38
rs753013549 Chr.22: 19851332-19851334 on Build GRCh38
rs537083361 Chr.22: 19851357-19851357 on Build GRCh38
rs762201539 Chr.22: 19851452-19851452 on Build GRCh38
rs201085521 Chr.22: 19851392-19851392 on Build GRCh38
rs772856177 Chr.22: 19851382-19851382 on Build GRCh38
rs555274757 Chr.22: 19851427-19851427 on Build GRCh38
rs769500031 Chr.22: 19851431-19851431 on Build GRCh38
rs1019262497 Chr.22: 19851354-19851354 on Build GRCh38
rs139112255 Chr.22: 19851399-19851399 on Build GRCh38
rs143002326 Chr.22: 19851496-19851496 on Build GRCh38
rs760147830 Chr.22: 19851436-19851436 on Build GRCh38
rs748380390 Chr.22: 19851511-19851511 on Build GRCh38
rs771017052 Chr.22: 19851329-19851329 on Build GRCh38
rs763737723 Chr.22: 19851351-19851351 on Build GRCh38

View Full Product Details