Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 61683788-61684013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRIP1
Entrez Gene ID
83990
Gene Name
BRCA1 interacting protein C-terminal helicase 1
Gene Aliases
BACH1, FANCJ, OF
UniGene
Hs.128903
Gene Chromosome Location
Chr.17: 61679186-61864120 on Build GRCh38
Transcripts
NM_032043.2 XM_011525332.2 XM_011525333.2 XM_011525334.2 XM_011525335.2 XM_011525336.2 XM_011525337.2 XM_011525338.2 XM_017025200.1 XM_017025201.1 XM_017025202.1 XM_017025203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7230322 c.3094A>T p.S1032C Chr.17: 61683952-61683952 on Build GRCh38
562079 c.3070G>T p.G1024W Chr.17: 61683976-61683976 on Build GRCh38
5386963 c.3123G>A p.M1041I Chr.17: 61683923-61683923 on Build GRCh38
5386961 c.3254G>A p.R1085K Chr.17: 61683792-61683792 on Build GRCh38
5386965 c.3067C>A p.L1023I Chr.17: 61683979-61683979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767255426 Chr.17: 61683957-61683957 on Build GRCh38
rs763162379 Chr.17: 61683950-61683950 on Build GRCh38
rs772507914 Chr.17: 61683901-61683901 on Build GRCh38
rs45437094 Chr.17: 61683943-61683943 on Build GRCh38
rs769310105 Chr.17: 61683929-61683929 on Build GRCh38
rs45540240 Chr.17: 61683851-61683851 on Build GRCh38
rs777213170 Chr.17: 61683837-61683837 on Build GRCh38
rs761225576 Chr.17: 61683924-61683924 on Build GRCh38
rs371185409 Chr.17: 61683967-61683967 on Build GRCh38
rs755949409 Chr.17: 61683960-61683960 on Build GRCh38
rs747345595 Chr.17: 61683978-61683978 on Build GRCh38
rs864622113 Chr.17: 61683819-61683819 on Build GRCh38
rs776002434 Chr.17: 61683922-61683922 on Build GRCh38
rs878855153 Chr.17: 61683847-61683847 on Build GRCh38
rs786201657 Chr.17: 61683968-61683968 on Build GRCh38
rs587782808 Chr.17: 61683982-61683982 on Build GRCh38
rs575998972 Chr.17: 61683858-61683858 on Build GRCh38
rs863224804 Chr.17: 61683966-61683966 on Build GRCh38
rs368867532 Chr.17: 61683823-61683823 on Build GRCh38
rs730881645 Chr.17: 61683850-61683850 on Build GRCh38
rs149016505 Chr.17: 61683868-61683868 on Build GRCh38
rs896510433 Chr.17: 61683903-61683903 on Build GRCh38
rs876659119 Chr.17: 61683949-61683949 on Build GRCh38
rs367816363 Chr.17: 61683942-61683942 on Build GRCh38
rs756074244 Chr.17: 61683832-61683832 on Build GRCh38
rs786204068 Chr.17: 61683831-61683831 on Build GRCh38
rs147119272 Chr.17: 61683976-61683976 on Build GRCh38
rs786203344 Chr.17: 61683906-61683906 on Build GRCh38
rs748598593 Chr.17: 61683838-61683838 on Build GRCh38
rs756712872 Chr.17: 61683970-61683970 on Build GRCh38
rs773433456 Chr.17: 61683960-61683960 on Build GRCh38
rs876659428 Chr.17: 61683928-61683928 on Build GRCh38
rs61754142 Chr.17: 61683977-61683977 on Build GRCh38
rs951001849 Chr.17: 61683876-61683876 on Build GRCh38
rs778430337 Chr.17: 61683865-61683865 on Build GRCh38
rs916937983 Chr.17: 61683855-61683855 on Build GRCh38
rs937490699 Chr.17: 61683953-61683953 on Build GRCh38
rs373040333 Chr.17: 61683897-61683897 on Build GRCh38
rs778500245 Chr.17: 61683974-61683974 on Build GRCh38
rs777660106 Chr.17: 61683965-61683965 on Build GRCh38
rs202228407 Chr.17: 61683947-61683947 on Build GRCh38
rs587781744 Chr.17: 61683975-61683975 on Build GRCh38
rs878855152 Chr.17: 61683857-61683857 on Build GRCh38
rs183928474 Chr.17: 61683822-61683822 on Build GRCh38
rs765416041 Chr.17: 61683946-61683946 on Build GRCh38
rs770517912 Chr.17: 61683861-61683861 on Build GRCh38

View Full Product Details