Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61802668-61802941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS1
Entrez Gene ID
3992
Gene Name
fatty acid desaturase 1
Gene Aliases
D5D, FADS6, FADSD5, LLCDL1, TU12
UniGene
Hs.503546
Gene Chromosome Location
Chr.11: 61799625-61817057 on Build GRCh38
Transcripts
NM_013402.4 XM_011545022.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4433980 c.1247C>T p.S416F Chr.11: 61802837-61802837 on Build GRCh38
5373025 c.1178G>T p.R393L Chr.11: 61802906-61802906 on Build GRCh38
6320123 c.1281C>A p.I427I Chr.11: 61802803-61802803 on Build GRCh38
5999506 c.1272C>T p.A424A Chr.11: 61802812-61802812 on Build GRCh38
929524 c.1241A>G p.Y414C Chr.11: 61802843-61802843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755117796 Chr.11: 61802709-61802711 on Build GRCh38
rs764353733 Chr.11: 61802836-61802836 on Build GRCh38
rs756750974 Chr.11: 61802896-61802896 on Build GRCh38
rs771890008 Chr.11: 61802784-61802784 on Build GRCh38
rs546978670 Chr.11: 61802704-61802704 on Build GRCh38
rs778652269 Chr.11: 61802770-61802770 on Build GRCh38
rs1013148394 Chr.11: 61802715-61802715 on Build GRCh38
rs781733916 Chr.11: 61802763-61802763 on Build GRCh38
rs922645035 Chr.11: 61802743-61802743 on Build GRCh38
rs747796745 Chr.11: 61802775-61802775 on Build GRCh38
rs748708405 Chr.11: 61802769-61802769 on Build GRCh38
rs767846209 Chr.11: 61802886-61802886 on Build GRCh38
rs374272165 Chr.11: 61802789-61802789 on Build GRCh38
rs964509278 Chr.11: 61802714-61802714 on Build GRCh38
rs115713429 Chr.11: 61802914-61802914 on Build GRCh38
rs759473846 Chr.11: 61802815-61802815 on Build GRCh38
rs762270502 Chr.11: 61802881-61802881 on Build GRCh38
rs770730137 Chr.11: 61802812-61802812 on Build GRCh38
rs746843286 Chr.11: 61802792-61802792 on Build GRCh38
rs776632991 Chr.11: 61802814-61802814 on Build GRCh38
rs748247775 Chr.11: 61802830-61802830 on Build GRCh38
rs757687195 Chr.11: 61802754-61802754 on Build GRCh38
rs539945596 Chr.11: 61802717-61802717 on Build GRCh38
rs934021221 Chr.11: 61802858-61802858 on Build GRCh38
rs146848923 Chr.11: 61802719-61802719 on Build GRCh38
rs777408109 Chr.11: 61802786-61802786 on Build GRCh38
rs769857419 Chr.11: 61802696-61802696 on Build GRCh38
rs769854507 Chr.11: 61802827-61802827 on Build GRCh38
rs201616473 Chr.11: 61802895-61802895 on Build GRCh38
rs879094043 Chr.11: 61802721-61802721 on Build GRCh38
rs751822479 Chr.11: 61802878-61802878 on Build GRCh38
rs200604746 Chr.11: 61802915-61802915 on Build GRCh38
rs372814217 Chr.11: 61802907-61802907 on Build GRCh38

View Full Product Details