Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45097299-45097562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUOX2
Entrez Gene ID
50506
Gene Name
dual oxidase 2
Gene Aliases
LNOX2, NOXEF2, P138-TOX, TDH6, THOX2
UniGene
Hs.71377
Gene Chromosome Location
Chr.15: 45092653-45114161 on Build GRCh38
Transcripts
NM_014080.4 XM_005254421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7114146 c.3720G>C p.L1240L Chr.15: 45097365-45097365 on Build GRCh38
1478154 c.3699C>T p.I1233I Chr.15: 45097386-45097386 on Build GRCh38
6650200 c.3758C>T p.P1253L Chr.15: 45097327-45097327 on Build GRCh38
3887050 c.3738C>T p.F1246F Chr.15: 45097347-45097347 on Build GRCh38
5732921 c.3761C>G p.A1254G Chr.15: 45097324-45097324 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751135600 Chr.15: 45097329-45097329 on Build GRCh38
rs374409729 Chr.15: 45097428-45097428 on Build GRCh38
rs769882380 Chr.15: 45097362-45097362 on Build GRCh38
rs753507689 Chr.15: 45097402-45097402 on Build GRCh38
rs748427684 Chr.15: 45097442-45097442 on Build GRCh38
rs530513960 Chr.15: 45097351-45097351 on Build GRCh38
rs372385025 Chr.15: 45097443-45097443 on Build GRCh38
rs549283819 Chr.15: 45097444-45097444 on Build GRCh38
rs761844122 Chr.15: 45097357-45097357 on Build GRCh38
rs964983021 Chr.15: 45097474-45097474 on Build GRCh38
rs759488791 Chr.15: 45097384-45097384 on Build GRCh38
rs756757978 Chr.15: 45097415-45097415 on Build GRCh38
rs528104610 Chr.15: 45097325-45097325 on Build GRCh38
rs371311874 Chr.15: 45097399-45097399 on Build GRCh38
rs906118758 Chr.15: 45097365-45097365 on Build GRCh38
rs897181270 Chr.15: 45097436-45097436 on Build GRCh38
rs1016778617 Chr.15: 45097368-45097368 on Build GRCh38
rs201697136 Chr.15: 45097439-45097439 on Build GRCh38
rs778518574 Chr.15: 45097426-45097426 on Build GRCh38
rs752492430 Chr.15: 45097400-45097400 on Build GRCh38
rs772669214 Chr.15: 45097327-45097327 on Build GRCh38
rs566008009 Chr.15: 45097476-45097476 on Build GRCh38
rs747431782 Chr.15: 45097437-45097437 on Build GRCh38
rs957231256 Chr.15: 45097473-45097473 on Build GRCh38
rs113832134 Chr.15: 45097441-45097441 on Build GRCh38
rs557823996 Chr.15: 45097500-45097500 on Build GRCh38
rs761011266 Chr.15: 45097369-45097369 on Build GRCh38
rs760088453 Chr.15: 45097377-45097379 on Build GRCh38
rs760457544 Chr.15: 45097401-45097401 on Build GRCh38
rs879197577 Chr.15: 45097393-45097393 on Build GRCh38
rs767954434 Chr.15: 45097381-45097399 on Build GRCh38
rs758989472 Chr.15: 45097433-45097433 on Build GRCh38
rs535018423 Chr.15: 45097477-45097477 on Build GRCh38
rs780644806 Chr.15: 45097435-45097435 on Build GRCh38
rs766871326 Chr.15: 45097373-45097374 on Build GRCh38
rs769007578 Chr.15: 45097343-45097343 on Build GRCh38
rs879081985 Chr.15: 45097396-45097396 on Build GRCh38
rs879006855 Chr.15: 45097397-45097397 on Build GRCh38
rs780589999 Chr.15: 45097331-45097331 on Build GRCh38
rs200910210 Chr.15: 45097333-45097333 on Build GRCh38
rs140663764 Chr.15: 45097326-45097326 on Build GRCh38
rs755518311 Chr.15: 45097410-45097410 on Build GRCh38
rs774959435 Chr.15: 45097374-45097374 on Build GRCh38

View Full Product Details