Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48939396-48939659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHDH
Entrez Gene ID
27294
Gene Name
dihydrodiol dehydrogenase
Gene Aliases
2DD, HUM2DD
UniGene
Hs.631555
Gene Chromosome Location
Chr.19: 48932790-48944969 on Build GRCh38
Transcripts
NM_014475.3 XM_005258748.4 XM_017026598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6643261 c.379C>A p.R127S Chr.19: 48939461-48939461 on Build GRCh38
1203504 c.380G>A p.R127H Chr.19: 48939462-48939462 on Build GRCh38
5507675 c.419C>T p.A140V Chr.19: 48939501-48939501 on Build GRCh38
3757028 c.396C>T p.S132S Chr.19: 48939478-48939478 on Build GRCh38
6643263 c.555C>T p.T185T Chr.19: 48939637-48939637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779901742 Chr.19: 48939621-48939621 on Build GRCh38
rs773323241 Chr.19: 48939530-48939530 on Build GRCh38
rs756827825 Chr.19: 48939569-48939569 on Build GRCh38
rs543413361 Chr.19: 48939600-48939600 on Build GRCh38
rs768398076 Chr.19: 48939627-48939627 on Build GRCh38
rs775768762 Chr.19: 48939593-48939593 on Build GRCh38
rs746802676 Chr.19: 48939624-48939624 on Build GRCh38
rs374671513 Chr.19: 48939469-48939469 on Build GRCh38
rs780647541 Chr.19: 48939440-48939440 on Build GRCh38
rs765185031 Chr.19: 48939526-48939526 on Build GRCh38
rs143910604 Chr.19: 48939598-48939598 on Build GRCh38
rs774339155 Chr.19: 48939494-48939494 on Build GRCh38
rs150679060 Chr.19: 48939581-48939581 on Build GRCh38
rs865845501 Chr.19: 48939461-48939461 on Build GRCh38
rs771831984 Chr.19: 48939577-48939577 on Build GRCh38
rs397960489 Chr.19: 48939599-48939600 on Build GRCh38
rs888244591 Chr.19: 48939513-48939513 on Build GRCh38
rs765758476 Chr.19: 48939598-48939599 on Build GRCh38
rs951504808 Chr.19: 48939594-48939594 on Build GRCh38
rs748796466 Chr.19: 48939486-48939486 on Build GRCh38
rs777340631 Chr.19: 48939563-48939563 on Build GRCh38
rs542046418 Chr.19: 48939552-48939552 on Build GRCh38
rs745648911 Chr.19: 48939501-48939501 on Build GRCh38
rs868661850 Chr.19: 48939503-48939503 on Build GRCh38
rs1018435252 Chr.19: 48939521-48939521 on Build GRCh38
rs984091958 Chr.19: 48939597-48939597 on Build GRCh38
rs751945250 Chr.19: 48939426-48939426 on Build GRCh38
rs752121060 Chr.19: 48939547-48939547 on Build GRCh38
rs373621522 Chr.19: 48939570-48939570 on Build GRCh38
rs2270940 Chr.19: 48939478-48939478 on Build GRCh38
rs140112768 Chr.19: 48939533-48939533 on Build GRCh38
rs377671385 Chr.19: 48939574-48939574 on Build GRCh38
rs938502581 Chr.19: 48939479-48939479 on Build GRCh38
rs766992335 Chr.19: 48939541-48939541 on Build GRCh38
rs766325147 Chr.19: 48939591-48939592 on Build GRCh38
rs142917861 Chr.19: 48939575-48939575 on Build GRCh38
rs554003656 Chr.19: 48939554-48939554 on Build GRCh38
rs758323302 Chr.19: 48939618-48939618 on Build GRCh38
rs113857438 Chr.19: 48939565-48939565 on Build GRCh38
rs148646797 Chr.19: 48939562-48939562 on Build GRCh38
rs763168931 Chr.19: 48939534-48939534 on Build GRCh38
rs371726493 Chr.19: 48939462-48939462 on Build GRCh38
rs750319188 Chr.19: 48939614-48939614 on Build GRCh38
rs112021846 Chr.19: 48939524-48939524 on Build GRCh38
rs746817769 Chr.19: 48939576-48939576 on Build GRCh38
rs760854899 Chr.19: 48939489-48939489 on Build GRCh38
rs753494372 Chr.19: 48939611-48939611 on Build GRCh38
rs1006726849 Chr.19: 48939599-48939599 on Build GRCh38
rs78637763 Chr.19: 48939599-48939599 on Build GRCh38
rs200486505 Chr.19: 48939631-48939631 on Build GRCh38
rs753333350 Chr.19: 48939564-48939564 on Build GRCh38
rs774953544 Chr.19: 48939588-48939588 on Build GRCh38
rs765624585 Chr.19: 48939549-48939549 on Build GRCh38
rs756697763 Chr.19: 48939613-48939613 on Build GRCh38
rs745818274 Chr.19: 48939571-48939571 on Build GRCh38
rs755465801 Chr.19: 48939435-48939435 on Build GRCh38
rs980943242 Chr.19: 48939580-48939580 on Build GRCh38
rs1056917899 Chr.19: 48939508-48939508 on Build GRCh38
rs546059210 Chr.19: 48939582-48939582 on Build GRCh38
rs3830420 Chr.19: 48939592-48939593 on Build GRCh38
rs139938386 Chr.19: 48939587-48939587 on Build GRCh38
rs770823489 Chr.19: 48939489-48939489 on Build GRCh38
rs762127594 Chr.19: 48939525-48939525 on Build GRCh38
rs34888386 Chr.19: 48939518-48939518 on Build GRCh38

View Full Product Details