Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22067400-22067639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMTN
Entrez Gene ID
2039
Gene Name
dematin actin binding protein
Gene Aliases
DMT, EPB49
UniGene
Hs.106124
Gene Chromosome Location
Chr.8: 22048955-22082527 on Build GRCh38
Transcripts
NM_001114135.4 NM_001114136.2 NM_001114137.3 NM_001114138.2 NM_001114139.3 NM_001302816.2 NM_001302817.2 NM_001323378.1 NM_001323380.1 NM_001323381.1 NM_001323382.1 NM_001323383.1 NM_001323384.1 NM_001323385.1 NM_001323387.1 NM_001323388.1 NM_001323389.1 NM_001323390.1 NM_001323391.1 NM_001323392.1 NM_001323393.1 NM_001323394.1 NM_001323395.1 NM_001323396.1 NM_001323397.1 NM_001323398.1 NM_001323399.1 NM_001323400.1 NM_001323401.1 NM_001978.4 XM_005273432.1 XM_005273433.1 XM_005273442.1 XM_011544435.1 XM_017013189.1 XM_017013190.1 XM_017013191.1 XM_017013192.1 XM_017013193.1 XM_017013194.1 XM_017013195.1 XM_017013196.1 XM_017013197.1 XM_017013198.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4458809 c.109C>T p.Q37* Chr.8: 22067542-22067542 on Build GRCh38
1456019 c.168C>T p.I56I Chr.8: 22067601-22067601 on Build GRCh38
5501905 c.155C>A p.A52D Chr.8: 22067588-22067588 on Build GRCh38
4414943 c.159C>T p.I53I Chr.8: 22067592-22067592 on Build GRCh38
4458808 c.109C>T p.Q37* Chr.8: 22067542-22067542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555616444 Chr.8: 22067568-22067568 on Build GRCh38
rs761947142 Chr.8: 22067601-22067601 on Build GRCh38
rs11135823 Chr.8: 22067433-22067433 on Build GRCh38
rs953532917 Chr.8: 22067451-22067451 on Build GRCh38
rs569856045 Chr.8: 22067465-22067465 on Build GRCh38
rs911934108 Chr.8: 22067428-22067428 on Build GRCh38
rs757555781 Chr.8: 22067498-22067498 on Build GRCh38
rs56171291 Chr.8: 22067459-22067459 on Build GRCh38
rs947894684 Chr.8: 22067616-22067616 on Build GRCh38
rs770590806 Chr.8: 22067546-22067546 on Build GRCh38
rs763373164 Chr.8: 22067506-22067506 on Build GRCh38
rs779230709 Chr.8: 22067508-22067508 on Build GRCh38
rs151041134 Chr.8: 22067509-22067509 on Build GRCh38
rs771873401 Chr.8: 22067551-22067551 on Build GRCh38
rs774015077 Chr.8: 22067548-22067548 on Build GRCh38
rs534322027 Chr.8: 22067593-22067593 on Build GRCh38
rs760683083 Chr.8: 22067581-22067581 on Build GRCh38
rs1042628939 Chr.8: 22067454-22067454 on Build GRCh38
rs992079429 Chr.8: 22067599-22067599 on Build GRCh38
rs748736580 Chr.8: 22067545-22067545 on Build GRCh38
rs764053941 Chr.8: 22067583-22067583 on Build GRCh38
rs764252247 Chr.8: 22067480-22067480 on Build GRCh38
rs747538366 Chr.8: 22067522-22067522 on Build GRCh38
rs758762092 Chr.8: 22067511-22067511 on Build GRCh38
rs538701037 Chr.8: 22067435-22067435 on Build GRCh38
rs769135442 Chr.8: 22067502-22067502 on Build GRCh38
rs546719622 Chr.8: 22067432-22067432 on Build GRCh38
rs754177240 Chr.8: 22067492-22067492 on Build GRCh38
rs372864638 Chr.8: 22067540-22067540 on Build GRCh38
rs771271089 Chr.8: 22067515-22067515 on Build GRCh38
rs140385917 Chr.8: 22067592-22067592 on Build GRCh38
rs139466873 Chr.8: 22067528-22067528 on Build GRCh38

View Full Product Details