Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 31071602-31071845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTHL17
Entrez Gene ID
53940
Gene Name
ferritin heavy chain like 17
Gene Aliases
CT38
UniGene
Hs.333125
Gene Chromosome Location
Chr.X: 31071241-31072053 on Build GRCh38
Transcripts
NM_031894.2

SNP Detail

SNP ID Chromosome Location
rs748349892 Chr.X: 31071816-31071816 on Build GRCh38
rs764865798 Chr.X: 31071740-31071742 on Build GRCh38
rs138241353 Chr.X: 31071815-31071815 on Build GRCh38
rs758861598 Chr.X: 31071659-31071659 on Build GRCh38
rs753817162 Chr.X: 31071805-31071805 on Build GRCh38
rs367975849 Chr.X: 31071761-31071761 on Build GRCh38
rs777245976 Chr.X: 31071804-31071804 on Build GRCh38
rs551562864 Chr.X: 31071823-31071823 on Build GRCh38
rs759887888 Chr.X: 31071754-31071754 on Build GRCh38
rs894705242 Chr.X: 31071702-31071702 on Build GRCh38
rs2159525 Chr.X: 31071732-31071732 on Build GRCh38
rs200675236 Chr.X: 31071681-31071681 on Build GRCh38
rs747326164 Chr.X: 31071695-31071695 on Build GRCh38
rs200714507 Chr.X: 31071653-31071653 on Build GRCh38
rs762437942 Chr.X: 31071623-31071623 on Build GRCh38
rs438870 Chr.X: 31071813-31071813 on Build GRCh38
rs769951116 Chr.X: 31071716-31071716 on Build GRCh38
rs374535093 Chr.X: 31071645-31071645 on Build GRCh38
rs200069856 Chr.X: 31071686-31071686 on Build GRCh38
rs370626340 Chr.X: 31071666-31071666 on Build GRCh38
rs761801390 Chr.X: 31071714-31071714 on Build GRCh38
rs755602895 Chr.X: 31071790-31071790 on Build GRCh38
rs776852674 Chr.X: 31071710-31071710 on Build GRCh38
rs752354910 Chr.X: 31071774-31071774 on Build GRCh38
rs143159310 Chr.X: 31071725-31071725 on Build GRCh38
rs140427727 Chr.X: 31071811-31071811 on Build GRCh38
rs73617043 Chr.X: 31071756-31071756 on Build GRCh38
rs747627045 Chr.X: 31071679-31071679 on Build GRCh38
rs201587622 Chr.X: 31071744-31071744 on Build GRCh38
rs899256338 Chr.X: 31071715-31071715 on Build GRCh38
rs751965888 Chr.X: 31071728-31071728 on Build GRCh38
rs147611025 Chr.X: 31071660-31071660 on Build GRCh38
rs757185333 Chr.X: 31071809-31071809 on Build GRCh38
rs749175316 Chr.X: 31071684-31071684 on Build GRCh38
rs754554679 Chr.X: 31071676-31071676 on Build GRCh38
rs755586266 Chr.X: 31071680-31071680 on Build GRCh38
rs745659539 Chr.X: 31071812-31071812 on Build GRCh38
rs751305567 Chr.X: 31071665-31071665 on Build GRCh38
rs61739942 Chr.X: 31071724-31071724 on Build GRCh38
rs943397335 Chr.X: 31071663-31071663 on Build GRCh38
rs780819993 Chr.X: 31071677-31071677 on Build GRCh38
rs768751338 Chr.X: 31071698-31071698 on Build GRCh38
rs773874373 Chr.X: 31071722-31071722 on Build GRCh38

View Full Product Details