Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75498571-75498832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIPR1
Entrez Gene ID
11010
Gene Name
GLI pathogenesis related 1
Gene Aliases
CRISP7, GLIPR, RTVP1
UniGene
Hs.205558
Gene Chromosome Location
Chr.12: 75480733-75501936 on Build GRCh38
Transcripts
NM_006851.2 XM_011537775.2 XM_017018721.1 NM_007043.6

Gene Symbol
KRR1
Entrez Gene ID
11103
Gene Name
KRR1, small subunit processome component homolog
Gene Aliases
HRB2, RIP-1
UniGene
Hs.645517
Gene Chromosome Location
Chr.12: 75497639-75511638 on Build GRCh38
Transcripts
NM_006851.2 XM_011537775.2 XM_017018721.1 NM_007043.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5361768 c.627A>T p.R209R Chr.12: 75498701-75498701 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200482879 Chr.12: 75498682-75498682 on Build GRCh38
rs3736391 Chr.12: 75498700-75498700 on Build GRCh38
rs749486931 Chr.12: 75498683-75498684 on Build GRCh38
rs775792426 Chr.12: 75498654-75498654 on Build GRCh38
rs759126841 Chr.12: 75498648-75498648 on Build GRCh38
rs779519062 Chr.12: 75498658-75498662 on Build GRCh38
rs946401113 Chr.12: 75498680-75498680 on Build GRCh38
rs780397471 Chr.12: 75498685-75498685 on Build GRCh38
rs764414512 Chr.12: 75498660-75498660 on Build GRCh38
rs148540028 Chr.12: 75498628-75498629 on Build GRCh38
rs910832176 Chr.12: 75498689-75498689 on Build GRCh38
rs751881802 Chr.12: 75498692-75498692 on Build GRCh38
rs564929541 Chr.12: 75498735-75498736 on Build GRCh38
rs371118659 Chr.12: 75498772-75498772 on Build GRCh38
rs777493094 Chr.12: 75498699-75498699 on Build GRCh38
rs199692388 Chr.12: 75498720-75498720 on Build GRCh38
rs751011510 Chr.12: 75498744-75498744 on Build GRCh38
rs201786239 Chr.12: 75498724-75498724 on Build GRCh38
rs773631187 Chr.12: 75498683-75498683 on Build GRCh38
rs144968885 Chr.12: 75498701-75498701 on Build GRCh38
rs1010257734 Chr.12: 75498787-75498787 on Build GRCh38
rs373895623 Chr.12: 75498702-75498702 on Build GRCh38
rs755385277 Chr.12: 75498642-75498642 on Build GRCh38
rs201199874 Chr.12: 75498659-75498659 on Build GRCh38
rs914892418 Chr.12: 75498662-75498663 on Build GRCh38
rs781048709 Chr.12: 75498681-75498683 on Build GRCh38
rs763107449 Chr.12: 75498753-75498753 on Build GRCh38
rs777563404 Chr.12: 75498800-75498800 on Build GRCh38
rs905074546 Chr.12: 75498798-75498798 on Build GRCh38
rs201806603 Chr.12: 75498725-75498725 on Build GRCh38
rs771181014 Chr.12: 75498711-75498712 on Build GRCh38
rs771733392 Chr.12: 75498649-75498649 on Build GRCh38
rs200107199 Chr.12: 75498732-75498732 on Build GRCh38
rs765695668 Chr.12: 75498738-75498738 on Build GRCh38
rs755963187 Chr.12: 75498697-75498697 on Build GRCh38
rs751829481 Chr.12: 75498791-75498791 on Build GRCh38
rs1049665404 Chr.12: 75498620-75498620 on Build GRCh38
rs773735837 Chr.12: 75498646-75498646 on Build GRCh38
rs765304088 Chr.12: 75498666-75498666 on Build GRCh38
rs757944016 Chr.12: 75498647-75498648 on Build GRCh38
rs766627075 Chr.12: 75498788-75498788 on Build GRCh38
rs375418899 Chr.12: 75498794-75498794 on Build GRCh38
rs3736392 Chr.12: 75498706-75498706 on Build GRCh38
rs569157020 Chr.12: 75498663-75498663 on Build GRCh38
rs747996513 Chr.12: 75498655-75498655 on Build GRCh38
rs747843506 Chr.12: 75498681-75498682 on Build GRCh38
rs776874596 Chr.12: 75498729-75498729 on Build GRCh38
rs768440812 Chr.12: 75498670-75498671 on Build GRCh38
rs374132123 Chr.12: 75498713-75498713 on Build GRCh38
rs760377605 Chr.12: 75498721-75498721 on Build GRCh38
rs932566120 Chr.12: 75498608-75498608 on Build GRCh38
rs1022969224 Chr.12: 75498741-75498741 on Build GRCh38
rs762232836 Chr.12: 75498664-75498664 on Build GRCh38
rs769703066 Chr.12: 75498682-75498683 on Build GRCh38
rs535834784 Chr.12: 75498661-75498661 on Build GRCh38
rs1049203286 Chr.12: 75498714-75498714 on Build GRCh38
rs750492037 Chr.12: 75498671-75498671 on Build GRCh38
rs375247979 Chr.12: 75498679-75498679 on Build GRCh38

View Full Product Details