Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24225196-24225455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT5
Entrez Gene ID
2687
Gene Name
gamma-glutamyltransferase 5
Gene Aliases
GGL, GGT 5, GGT-REL, GGTLA1
UniGene
Hs.437156
Gene Chromosome Location
Chr.22: 24219654-24245142 on Build GRCh38
Transcripts
NM_001099781.2 NM_001099782.2 NM_001302464.1 NM_001302465.1 NM_004121.3 XM_005261557.2 XM_005261558.2 XM_011530133.1 XM_011530134.1 XM_011530135.1 XM_011530136.1 XM_011530137.2 XM_017028768.1 XM_017028769.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4399676 c.1466G>A p.G489E Chr.22: 24225282-24225282 on Build GRCh38
5057727 c.1446_1447insG p.S483fs*30 Chr.22: 24225301-24225302 on Build GRCh38
4164902 c.1430T>A p.I477N Chr.22: 24225318-24225318 on Build GRCh38
6891914 c.1410C>T p.S470S Chr.22: 24225338-24225338 on Build GRCh38
4164901 c.1431C>T p.I477I Chr.22: 24225317-24225317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777898121 Chr.22: 24225337-24225337 on Build GRCh38
rs1041111035 Chr.22: 24225304-24225304 on Build GRCh38
rs753628065 Chr.22: 24225411-24225411 on Build GRCh38
rs150898918 Chr.22: 24225284-24225284 on Build GRCh38
rs765588510 Chr.22: 24225263-24225263 on Build GRCh38
rs528779081 Chr.22: 24225303-24225303 on Build GRCh38
rs374329258 Chr.22: 24225433-24225433 on Build GRCh38
rs139383582 Chr.22: 24225299-24225299 on Build GRCh38
rs755737041 Chr.22: 24225388-24225388 on Build GRCh38
rs777095964 Chr.22: 24225223-24225223 on Build GRCh38
rs200684960 Chr.22: 24225222-24225222 on Build GRCh38
rs752800899 Chr.22: 24225323-24225324 on Build GRCh38
rs542440435 Chr.22: 24225396-24225396 on Build GRCh38
rs903249652 Chr.22: 24225405-24225405 on Build GRCh38
rs866363144 Chr.22: 24225270-24225270 on Build GRCh38
rs748071790 Chr.22: 24225421-24225421 on Build GRCh38
rs776035379 Chr.22: 24225431-24225431 on Build GRCh38
rs765401891 Chr.22: 24225257-24225258 on Build GRCh38
rs761249346 Chr.22: 24225379-24225379 on Build GRCh38
rs747032399 Chr.22: 24225342-24225342 on Build GRCh38
rs999205973 Chr.22: 24225265-24225265 on Build GRCh38
rs1030793569 Chr.22: 24225274-24225274 on Build GRCh38
rs201616574 Chr.22: 24225219-24225219 on Build GRCh38
rs776743050 Chr.22: 24225350-24225350 on Build GRCh38
rs759800101 Chr.22: 24225234-24225234 on Build GRCh38
rs751162471 Chr.22: 24225377-24225377 on Build GRCh38
rs771235441 Chr.22: 24225346-24225346 on Build GRCh38
rs55890517 Chr.22: 24225426-24225426 on Build GRCh38
rs892435899 Chr.22: 24225247-24225247 on Build GRCh38
rs985183445 Chr.22: 24225375-24225375 on Build GRCh38
rs529621965 Chr.22: 24225359-24225359 on Build GRCh38
rs746133651 Chr.22: 24225427-24225427 on Build GRCh38
rs1026801373 Chr.22: 24225351-24225351 on Build GRCh38
rs200146637 Chr.22: 24225356-24225356 on Build GRCh38
rs764663086 Chr.22: 24225370-24225370 on Build GRCh38
rs558318043 Chr.22: 24225428-24225428 on Build GRCh38
rs767011018 Chr.22: 24225382-24225382 on Build GRCh38
rs376709868 Chr.22: 24225369-24225369 on Build GRCh38
rs564711431 Chr.22: 24225295-24225295 on Build GRCh38
rs768572532 Chr.22: 24225331-24225331 on Build GRCh38
rs763495821 Chr.22: 24225283-24225283 on Build GRCh38
rs750040366 Chr.22: 24225300-24225300 on Build GRCh38
rs778801768 Chr.22: 24225416-24225416 on Build GRCh38
rs899927211 Chr.22: 24225317-24225317 on Build GRCh38
rs113295783 Chr.22: 24225243-24225243 on Build GRCh38
rs143616956 Chr.22: 24225383-24225383 on Build GRCh38
rs150033594 Chr.22: 24225302-24225302 on Build GRCh38
rs756973423 Chr.22: 24225296-24225296 on Build GRCh38
rs7288201 Chr.22: 24225325-24225325 on Build GRCh38
rs370492248 Chr.22: 24225275-24225275 on Build GRCh38
rs780434605 Chr.22: 24225412-24225412 on Build GRCh38
rs140052476 Chr.22: 24225355-24225355 on Build GRCh38

View Full Product Details