Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92832889-92833156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM69A
Entrez Gene ID
388650
Gene Name
family with sequence similarity 69 member A
Gene Aliases
-
UniGene
Hs.180946
Gene Chromosome Location
Chr.1: 92832729-92961522 on Build GRCh38
Transcripts
NM_001252273.1 NM_000969.3

Gene Symbol
RPL5
Entrez Gene ID
6125
Gene Name
ribosomal protein L5
Gene Aliases
DBA6, L5, MSTP030, PPP1R135
UniGene
Hs.532359
Gene Chromosome Location
Chr.1: 92832037-92841924 on Build GRCh38
Transcripts
NM_001252273.1 NM_000969.3

SNP Detail

SNP ID Chromosome Location
rs1038820871 Chr.1: 92832925-92832925 on Build GRCh38
rs749996738 Chr.1: 92833001-92833001 on Build GRCh38
rs538222041 Chr.1: 92832920-92832920 on Build GRCh38
rs554425454 Chr.1: 92833064-92833064 on Build GRCh38
rs944013709 Chr.1: 92833127-92833127 on Build GRCh38
rs752893229 Chr.1: 92832973-92832973 on Build GRCh38
rs746149734 Chr.1: 92833051-92833051 on Build GRCh38
rs776946966 Chr.1: 92832997-92832998 on Build GRCh38
rs780882370 Chr.1: 92832993-92832993 on Build GRCh38
rs748056308 Chr.1: 92833033-92833033 on Build GRCh38
rs933125743 Chr.1: 92833120-92833120 on Build GRCh38
rs769850716 Chr.1: 92833055-92833055 on Build GRCh38
rs778009669 Chr.1: 92832971-92832980 on Build GRCh38
rs948229949 Chr.1: 92833068-92833068 on Build GRCh38
rs763285257 Chr.1: 92832978-92832978 on Build GRCh38
rs764452993 Chr.1: 92832979-92832979 on Build GRCh38
rs773100754 Chr.1: 92833037-92833037 on Build GRCh38
rs914510645 Chr.1: 92833099-92833099 on Build GRCh38
rs901616013 Chr.1: 92832931-92832931 on Build GRCh38
rs368287683 Chr.1: 92832969-92832969 on Build GRCh38
rs538839176 Chr.1: 92832958-92832958 on Build GRCh38
rs754054992 Chr.1: 92832978-92832981 on Build GRCh38
rs748961192 Chr.1: 92833024-92833024 on Build GRCh38
rs138346863 Chr.1: 92833026-92833026 on Build GRCh38
rs763520381 Chr.1: 92833118-92833118 on Build GRCh38
rs748638969 Chr.1: 92833042-92833042 on Build GRCh38
rs372466975 Chr.1: 92833019-92833019 on Build GRCh38
rs775694391 Chr.1: 92833059-92833059 on Build GRCh38
rs145309245 Chr.1: 92833122-92833122 on Build GRCh38
rs369438439 Chr.1: 92833051-92833051 on Build GRCh38
rs751914143 Chr.1: 92833067-92833067 on Build GRCh38
rs943688624 Chr.1: 92833095-92833095 on Build GRCh38
rs755666517 Chr.1: 92833006-92833006 on Build GRCh38
rs112010150 Chr.1: 92832959-92832959 on Build GRCh38
rs139405785 Chr.1: 92833106-92833106 on Build GRCh38
rs978195062 Chr.1: 92832994-92832994 on Build GRCh38
rs556666585 Chr.1: 92832942-92832942 on Build GRCh38
rs143998608 Chr.1: 92833115-92833115 on Build GRCh38
rs943262792 Chr.1: 92833105-92833105 on Build GRCh38
rs772003988 Chr.1: 92833034-92833034 on Build GRCh38
rs542477366 Chr.1: 92833028-92833028 on Build GRCh38
rs1026005119 Chr.1: 92832952-92832952 on Build GRCh38
rs929986496 Chr.1: 92832918-92832918 on Build GRCh38
rs768233834 Chr.1: 92833078-92833078 on Build GRCh38
rs763178068 Chr.1: 92833061-92833061 on Build GRCh38
rs374703782 Chr.1: 92833088-92833088 on Build GRCh38
rs61634219 Chr.1: 92832943-92832943 on Build GRCh38
rs916284971 Chr.1: 92833060-92833060 on Build GRCh38
rs752021416 Chr.1: 92832984-92832984 on Build GRCh38
rs371750521 Chr.1: 92833079-92833079 on Build GRCh38
rs971532972 Chr.1: 92833035-92833035 on Build GRCh38

View Full Product Details