Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73241926-73242179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHDC1
Entrez Gene ID
80759
Gene Name
KH homology domain containing 1
Gene Aliases
C6orf147, C6orf148, Em:AC019205.8, NDG1, bA257K9.4
UniGene
Hs.433062
Gene Chromosome Location
Chr.6: 73241314-73310365 on Build GRCh38
Transcripts
NM_001251874.1 NM_030568.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
312240 c.293G>T p.R98L Chr.6: 73242057-73242057 on Build GRCh38
3162207 c.203G>A p.R68H Chr.6: 73242147-73242147 on Build GRCh38
4797808 c.273G>A p.Q91Q Chr.6: 73242077-73242077 on Build GRCh38
1445818 c.243G>T p.L81L Chr.6: 73242107-73242107 on Build GRCh38
5061934 c.264_265insTG p.G89fs*24 Chr.6: 73242085-73242086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372741660 Chr.6: 73242147-73242147 on Build GRCh38
rs759829191 Chr.6: 73242083-73242083 on Build GRCh38
rs756214465 Chr.6: 73242099-73242100 on Build GRCh38
rs757467678 Chr.6: 73242132-73242132 on Build GRCh38
rs746783459 Chr.6: 73242018-73242018 on Build GRCh38
rs754762273 Chr.6: 73242136-73242136 on Build GRCh38
rs199949364 Chr.6: 73242057-73242057 on Build GRCh38
rs768344998 Chr.6: 73242035-73242035 on Build GRCh38
rs564747296 Chr.6: 73241966-73241966 on Build GRCh38
rs750643533 Chr.6: 73242010-73242012 on Build GRCh38
rs988067195 Chr.6: 73242000-73242000 on Build GRCh38
rs569098928 Chr.6: 73242106-73242106 on Build GRCh38
rs773192554 Chr.6: 73242148-73242148 on Build GRCh38
rs766005212 Chr.6: 73242069-73242069 on Build GRCh38
rs181290286 Chr.6: 73242085-73242085 on Build GRCh38
rs546437325 Chr.6: 73241956-73241956 on Build GRCh38
rs780872192 Chr.6: 73242145-73242145 on Build GRCh38
rs535901829 Chr.6: 73242134-73242134 on Build GRCh38
rs17851969 Chr.6: 73242137-73242137 on Build GRCh38
rs925621462 Chr.6: 73242125-73242125 on Build GRCh38
rs918684169 Chr.6: 73241968-73241968 on Build GRCh38
rs572106838 Chr.6: 73242100-73242100 on Build GRCh38
rs748066811 Chr.6: 73242054-73242054 on Build GRCh38
rs532287244 Chr.6: 73241995-73241995 on Build GRCh38
rs780250371 Chr.6: 73242121-73242121 on Build GRCh38
rs774006126 Chr.6: 73242080-73242080 on Build GRCh38
rs369158621 Chr.6: 73242111-73242111 on Build GRCh38
rs768333060 Chr.6: 73242058-73242058 on Build GRCh38
rs779214439 Chr.6: 73242133-73242133 on Build GRCh38
rs576812355 Chr.6: 73242052-73242052 on Build GRCh38
rs762518328 Chr.6: 73242061-73242061 on Build GRCh38
rs753203314 Chr.6: 73242093-73242093 on Build GRCh38
rs770433424 Chr.6: 73242154-73242154 on Build GRCh38
rs916783826 Chr.6: 73242051-73242051 on Build GRCh38
rs925367878 Chr.6: 73241969-73241969 on Build GRCh38

View Full Product Details