Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30740569-30740830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLOT1
Entrez Gene ID
10211
Gene Name
flotillin 1
Gene Aliases
-
UniGene
Hs.179986
Gene Chromosome Location
Chr.6: 30727709-30742851 on Build GRCh38
Transcripts
NM_001318875.1 NM_005803.3 XM_005248780.3 XM_006714947.3 XM_017010157.1 XM_017010158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7102830 c.381C>G p.F127L Chr.6: 30740772-30740772 on Build GRCh38
4424929 c.428G>A p.G143D Chr.6: 30740725-30740725 on Build GRCh38
6349489 c.190-9C>G p.? Chr.6: 30740600-30740600 on Build GRCh38
4160576 c.377A>T p.K126I Chr.6: 30740776-30740776 on Build GRCh38
6349488 c.475-9C>G p.? Chr.6: 30740600-30740600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994333324 Chr.6: 30740782-30740782 on Build GRCh38
rs779461045 Chr.6: 30740748-30740748 on Build GRCh38
rs772245658 Chr.6: 30740768-30740768 on Build GRCh38
rs769056175 Chr.6: 30740747-30740747 on Build GRCh38
rs752498099 Chr.6: 30740653-30740653 on Build GRCh38
rs761072513 Chr.6: 30740809-30740811 on Build GRCh38
rs780865327 Chr.6: 30740709-30740709 on Build GRCh38
rs756789679 Chr.6: 30740694-30740694 on Build GRCh38
rs769166000 Chr.6: 30740634-30740634 on Build GRCh38
rs748240708 Chr.6: 30740754-30740754 on Build GRCh38
rs1041304126 Chr.6: 30740703-30740703 on Build GRCh38
rs776023752 Chr.6: 30740609-30740609 on Build GRCh38
rs749780693 Chr.6: 30740626-30740626 on Build GRCh38
rs761890901 Chr.6: 30740639-30740639 on Build GRCh38
rs962889608 Chr.6: 30740690-30740690 on Build GRCh38
rs527661312 Chr.6: 30740681-30740681 on Build GRCh38
rs755711731 Chr.6: 30740725-30740725 on Build GRCh38
rs762804018 Chr.6: 30740666-30740666 on Build GRCh38
rs1018437362 Chr.6: 30740665-30740665 on Build GRCh38
rs759456227 Chr.6: 30740644-30740644 on Build GRCh38
rs764039022 Chr.6: 30740687-30740687 on Build GRCh38
rs541987555 Chr.6: 30740715-30740715 on Build GRCh38
rs772477281 Chr.6: 30740640-30740640 on Build GRCh38
rs1050614467 Chr.6: 30740597-30740597 on Build GRCh38
rs773569412 Chr.6: 30740595-30740595 on Build GRCh38
rs139305218 Chr.6: 30740741-30740741 on Build GRCh38
rs774295690 Chr.6: 30740637-30740637 on Build GRCh38
rs901457596 Chr.6: 30740731-30740731 on Build GRCh38
rs770110434 Chr.6: 30740596-30740596 on Build GRCh38
rs12111310 Chr.6: 30740651-30740651 on Build GRCh38
rs751092275 Chr.6: 30740688-30740688 on Build GRCh38
rs749596091 Chr.6: 30740745-30740745 on Build GRCh38
rs766681940 Chr.6: 30740772-30740772 on Build GRCh38

View Full Product Details