Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31130623-31130878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSC1
Entrez Gene ID
1823
Gene Name
desmocollin 1
Gene Aliases
CDHF1, DG2/DG3
UniGene
Hs.567260
Gene Chromosome Location
Chr.18: 31129236-31162856 on Build GRCh38
Transcripts
NM_004948.3 NM_024421.2 NR_110785.1

Gene Symbol
DSCAS
Entrez Gene ID
101927698
Gene Name
DSC1/DSC2 antisense RNA
Gene Aliases
-
UniGene
Hs.579538
Gene Chromosome Location
Chr.18: 31101588-31162789 on Build GRCh38
Transcripts
NM_004948.3 NM_024421.2 NR_110785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7088550 c.2511G>C p.L837L Chr.18: 31130815-31130815 on Build GRCh38
5731307 c.2547G>A p.S849S Chr.18: 31130652-31130652 on Build GRCh38
4071745 c.2546C>T p.S849L Chr.18: 31130653-31130653 on Build GRCh38
3890872 c.2557G>A p.E853K Chr.18: 31130642-31130642 on Build GRCh38
1630559 c.2521T>C p.*841Q Chr.18: 31130805-31130805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768808857 Chr.18: 31130831-31130831 on Build GRCh38
rs775869885 Chr.18: 31130680-31130680 on Build GRCh38
rs987169867 Chr.18: 31130733-31130733 on Build GRCh38
rs753733385 Chr.18: 31130731-31130731 on Build GRCh38
rs761828107 Chr.18: 31130841-31130841 on Build GRCh38
rs1816634 Chr.18: 31130652-31130652 on Build GRCh38
rs774599033 Chr.18: 31130838-31130838 on Build GRCh38
rs879406769 Chr.18: 31130647-31130647 on Build GRCh38
rs746518527 Chr.18: 31130662-31130662 on Build GRCh38
rs777463222 Chr.18: 31130767-31130767 on Build GRCh38
rs771621552 Chr.18: 31130660-31130660 on Build GRCh38
rs746545856 Chr.18: 31130770-31130770 on Build GRCh38
rs1020138864 Chr.18: 31130736-31130736 on Build GRCh38
rs575190874 Chr.18: 31130819-31130819 on Build GRCh38
rs777167532 Chr.18: 31130672-31130672 on Build GRCh38
rs751647056 Chr.18: 31130708-31130708 on Build GRCh38
rs547250397 Chr.18: 31130745-31130746 on Build GRCh38
rs548099685 Chr.18: 31130816-31130817 on Build GRCh38
rs770973367 Chr.18: 31130775-31130777 on Build GRCh38
rs770289598 Chr.18: 31130817-31130817 on Build GRCh38
rs777762289 Chr.18: 31130739-31130740 on Build GRCh38
rs752347287 Chr.18: 31130745-31130745 on Build GRCh38
rs139389841 Chr.18: 31130673-31130673 on Build GRCh38
rs770236619 Chr.18: 31130847-31130848 on Build GRCh38
rs377023366 Chr.18: 31130723-31130723 on Build GRCh38
rs771394465 Chr.18: 31130665-31130665 on Build GRCh38
rs754736071 Chr.18: 31130653-31130653 on Build GRCh38
rs776881778 Chr.18: 31130782-31130783 on Build GRCh38
rs1006469113 Chr.18: 31130648-31130648 on Build GRCh38
rs538208950 Chr.18: 31130746-31130746 on Build GRCh38
rs763201484 Chr.18: 31130682-31130682 on Build GRCh38
rs200998108 Chr.18: 31130735-31130735 on Build GRCh38
rs374160616 Chr.18: 31130709-31130709 on Build GRCh38
rs985861 Chr.18: 31130656-31130656 on Build GRCh38
rs964620116 Chr.18: 31130792-31130792 on Build GRCh38
rs200960095 Chr.18: 31130661-31130661 on Build GRCh38
rs201404121 Chr.18: 31130674-31130674 on Build GRCh38
rs770236384 Chr.18: 31130679-31130679 on Build GRCh38
rs763397332 Chr.18: 31130821-31130821 on Build GRCh38
rs746321458 Chr.18: 31130812-31130812 on Build GRCh38
rs556534324 Chr.18: 31130786-31130786 on Build GRCh38
rs764373336 Chr.18: 31130685-31130685 on Build GRCh38
rs781771037 Chr.18: 31130806-31130806 on Build GRCh38
rs754787356 Chr.18: 31130734-31130734 on Build GRCh38
rs951402489 Chr.18: 31130732-31130732 on Build GRCh38

View Full Product Details