Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74929009-74929270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX5A
Entrez Gene ID
9377
Gene Name
cytochrome c oxidase subunit 5A
Gene Aliases
COX, COX-VA, VA
UniGene
Hs.401903
Gene Chromosome Location
Chr.15: 74920275-74938154 on Build GRCh38
Transcripts
NM_004255.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7233505 c.164G>A p.R55H Chr.15: 74929169-74929169 on Build GRCh38
4921222 c.172A>G p.T58A Chr.15: 74929161-74929161 on Build GRCh38
964940 c.115C>T p.R39C Chr.15: 74929218-74929218 on Build GRCh38
4935325 c.120C>G p.C40W Chr.15: 74929213-74929213 on Build GRCh38
4961835 c.213T>A p.R71R Chr.15: 74929120-74929120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575567611 Chr.15: 74929139-74929139 on Build GRCh38
rs766352622 Chr.15: 74929077-74929077 on Build GRCh38
rs775306455 Chr.15: 74929204-74929204 on Build GRCh38
rs375830469 Chr.15: 74929188-74929188 on Build GRCh38
rs1803316 Chr.15: 74929195-74929195 on Build GRCh38
rs912599455 Chr.15: 74929034-74929034 on Build GRCh38
rs780396486 Chr.15: 74929170-74929170 on Build GRCh38
rs746494422 Chr.15: 74929184-74929184 on Build GRCh38
rs372049492 Chr.15: 74929085-74929085 on Build GRCh38
rs751665674 Chr.15: 74929134-74929134 on Build GRCh38
rs749742231 Chr.15: 74929074-74929074 on Build GRCh38
rs201693225 Chr.15: 74929116-74929116 on Build GRCh38
rs759247989 Chr.15: 74929097-74929097 on Build GRCh38
rs900050999 Chr.15: 74929053-74929053 on Build GRCh38
rs150174803 Chr.15: 74929121-74929121 on Build GRCh38
rs774638701 Chr.15: 74929217-74929217 on Build GRCh38
rs546135039 Chr.15: 74929183-74929183 on Build GRCh38
rs372249618 Chr.15: 74929205-74929205 on Build GRCh38
rs766679274 Chr.15: 74929233-74929233 on Build GRCh38
rs200811470 Chr.15: 74929218-74929218 on Build GRCh38
rs542641042 Chr.15: 74929122-74929122 on Build GRCh38
rs138526218 Chr.15: 74929172-74929172 on Build GRCh38
rs755523610 Chr.15: 74929236-74929236 on Build GRCh38
rs768682367 Chr.15: 74929212-74929212 on Build GRCh38
rs200382369 Chr.15: 74929196-74929196 on Build GRCh38
rs140256516 Chr.15: 74929221-74929221 on Build GRCh38
rs200848101 Chr.15: 74929088-74929088 on Build GRCh38
rs757639315 Chr.15: 74929077-74929078 on Build GRCh38
rs143289611 Chr.15: 74929201-74929201 on Build GRCh38
rs764291064 Chr.15: 74929132-74929132 on Build GRCh38
rs750854500 Chr.15: 74929169-74929169 on Build GRCh38
rs954375210 Chr.15: 74929099-74929099 on Build GRCh38
rs761019932 Chr.15: 74929215-74929215 on Build GRCh38
rs780696817 Chr.15: 74929190-74929190 on Build GRCh38
rs780408288 Chr.15: 74929070-74929070 on Build GRCh38
rs754212993 Chr.15: 74929235-74929235 on Build GRCh38
rs11553377 Chr.15: 74929185-74929185 on Build GRCh38
rs777229415 Chr.15: 74929078-74929078 on Build GRCh38
rs767844945 Chr.15: 74929150-74929150 on Build GRCh38
rs760914811 Chr.15: 74929232-74929232 on Build GRCh38
rs779206976 Chr.15: 74929084-74929084 on Build GRCh38
rs112721341 Chr.15: 74929189-74929189 on Build GRCh38
rs1044532641 Chr.15: 74929108-74929108 on Build GRCh38
rs1016607099 Chr.15: 74929199-74929199 on Build GRCh38
rs1002612732 Chr.15: 74929123-74929123 on Build GRCh38
rs773308546 Chr.15: 74929231-74929231 on Build GRCh38
rs769828984 Chr.15: 74929111-74929111 on Build GRCh38

View Full Product Details