Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 86510750-86510975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXF1
Entrez Gene ID
2294
Gene Name
forkhead box F1
Gene Aliases
ACDMPV, FKHL5, FREAC1
UniGene
Hs.155591
Gene Chromosome Location
Chr.16: 86510527-86514464 on Build GRCh38
Transcripts
NM_001451.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3513090 c.183C>T p.I61I Chr.16: 86510752-86510752 on Build GRCh38
6560576 c.221T>C p.I74T Chr.16: 86510790-86510790 on Build GRCh38
5493476 c.202C>T p.R68C Chr.16: 86510771-86510771 on Build GRCh38
6666197 c.182G>A p.R61Q Chr.16: 86510826-86510826 on Build GRCh38
6523075 c.212T>A p.L71Q Chr.16: 86510781-86510781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199545626 Chr.16: 86510816-86510816 on Build GRCh38
rs121909336 Chr.16: 86510794-86510794 on Build GRCh38
rs775631156 Chr.16: 86510800-86510800 on Build GRCh38
rs767354703 Chr.16: 86510906-86510906 on Build GRCh38
rs916878606 Chr.16: 86510897-86510897 on Build GRCh38
rs757417468 Chr.16: 86510905-86510905 on Build GRCh38
rs779317458 Chr.16: 86510890-86510890 on Build GRCh38
rs1057518868 Chr.16: 86510849-86510849 on Build GRCh38
rs568384890 Chr.16: 86510778-86510778 on Build GRCh38
rs765587760 Chr.16: 86510797-86510797 on Build GRCh38
rs746392936 Chr.16: 86510860-86510861 on Build GRCh38
rs751560567 Chr.16: 86510879-86510879 on Build GRCh38
rs778760552 Chr.16: 86510939-86510939 on Build GRCh38
rs754649288 Chr.16: 86510918-86510918 on Build GRCh38
rs747736353 Chr.16: 86510771-86510771 on Build GRCh38
rs747596391 Chr.16: 86510945-86510945 on Build GRCh38
rs968174218 Chr.16: 86510811-86510811 on Build GRCh38
rs942303151 Chr.16: 86510780-86510780 on Build GRCh38
rs979593786 Chr.16: 86510824-86510824 on Build GRCh38
rs1031206293 Chr.16: 86510833-86510833 on Build GRCh38
rs202025999 Chr.16: 86510888-86510888 on Build GRCh38
rs370422274 Chr.16: 86510773-86510773 on Build GRCh38
rs111830706 Chr.16: 86510917-86510917 on Build GRCh38
rs545170184 Chr.16: 86510875-86510875 on Build GRCh38
rs771675021 Chr.16: 86510947-86510947 on Build GRCh38
rs764116889 Chr.16: 86510878-86510878 on Build GRCh38
rs927055364 Chr.16: 86510941-86510941 on Build GRCh38
rs201828578 Chr.16: 86510915-86510915 on Build GRCh38

View Full Product Details