Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 41223934-41224207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNB1
Entrez Gene ID
1499
Gene Name
catenin beta 1
Gene Aliases
CTNNB, MRD19, armadillo
UniGene
Hs.476018
Gene Chromosome Location
Chr.3: 41199451-41240448 on Build GRCh38
Transcripts
NM_001098209.1 NM_001098210.1 NM_001904.3 XM_005264886.2 XM_006712983.1 XM_006712984.1 XM_006712985.1 XM_017005738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7308134 c.8C>A p.T3N Chr.3: 41224076-41224076 on Build GRCh38
4117539 c.7A>G p.T3A Chr.3: 41224075-41224075 on Build GRCh38
6052286 c.13+60_241+11del624 p.? Chr.3: 41224141-41224764 on Build GRCh38
5966864 c.1_242-88del798 p.? Chr.3: 41223265-41224866 on Build GRCh38
27279 c.12A>T p.Q4H Chr.3: 41224080-41224080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535579990 Chr.3: 41224174-41224174 on Build GRCh38
rs1022004651 Chr.3: 41224139-41224139 on Build GRCh38
rs755744390 Chr.3: 41224031-41224031 on Build GRCh38
rs1023412374 Chr.3: 41224003-41224003 on Build GRCh38
rs968099699 Chr.3: 41224012-41224012 on Build GRCh38
rs1018043393 Chr.3: 41224046-41224046 on Build GRCh38
rs528470084 Chr.3: 41223999-41223999 on Build GRCh38
rs956534023 Chr.3: 41224069-41224069 on Build GRCh38
rs976344851 Chr.3: 41224062-41224062 on Build GRCh38
rs887847705 Chr.3: 41224173-41224173 on Build GRCh38
rs371316720 Chr.3: 41224086-41224086 on Build GRCh38
rs5743390 Chr.3: 41224060-41224060 on Build GRCh38
rs987832030 Chr.3: 41224134-41224134 on Build GRCh38
rs1005628566 Chr.3: 41224045-41224045 on Build GRCh38
rs534465078 Chr.3: 41224033-41224033 on Build GRCh38
rs1034809855 Chr.3: 41223979-41223979 on Build GRCh38
rs1047558623 Chr.3: 41224030-41224030 on Build GRCh38
rs569737786 Chr.3: 41224144-41224144 on Build GRCh38
rs749331498 Chr.3: 41224076-41224076 on Build GRCh38
rs754869050 Chr.3: 41224057-41224057 on Build GRCh38
rs4135376 Chr.3: 41224024-41224024 on Build GRCh38
rs1057389394 Chr.3: 41223961-41223961 on Build GRCh38
rs899703840 Chr.3: 41224049-41224049 on Build GRCh38
rs750034704 Chr.3: 41223995-41223995 on Build GRCh38
rs4135375 Chr.3: 41223971-41223971 on Build GRCh38
rs746112994 Chr.3: 41224112-41224112 on Build GRCh38
rs779188059 Chr.3: 41224101-41224101 on Build GRCh38
rs750102051 Chr.3: 41224019-41224019 on Build GRCh38
rs374696946 Chr.3: 41224175-41224175 on Build GRCh38
rs777295717 Chr.3: 41224127-41224127 on Build GRCh38
rs374922164 Chr.3: 41224092-41224092 on Build GRCh38
rs1029597775 Chr.3: 41224061-41224061 on Build GRCh38
rs990559334 Chr.3: 41224001-41224002 on Build GRCh38
rs967771421 Chr.3: 41224156-41224156 on Build GRCh38
rs960567648 Chr.3: 41223980-41223980 on Build GRCh38
rs775945275 Chr.3: 41224122-41224122 on Build GRCh38
rs751477759 Chr.3: 41224039-41224039 on Build GRCh38
rs549973660 Chr.3: 41224126-41224126 on Build GRCh38

View Full Product Details