Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74706276-74706521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRDL2
Entrez Gene ID
25884
Gene Name
chordin like 2
Gene Aliases
BNF1, CHL2
UniGene
Hs.432379
Gene Chromosome Location
Chr.11: 74696428-74731385 on Build GRCh38
Transcripts
NM_001278473.2 NM_001304390.1 NM_001304391.1 NM_001304415.1 NM_001304416.1 NM_001304417.1 NM_015424.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4036691 c.571C>A p.L191I Chr.11: 74706498-74706498 on Build GRCh38
1356964 c.570G>A p.S190S Chr.11: 74706499-74706499 on Build GRCh38
6628606 c.569C>T p.S190L Chr.11: 74706500-74706500 on Build GRCh38
4879844 c.573C>A p.L191L Chr.11: 74706496-74706496 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs367994435 Chr.11: 74706435-74706435 on Build GRCh38
rs924596155 Chr.11: 74706301-74706301 on Build GRCh38
rs1043441216 Chr.11: 74706438-74706438 on Build GRCh38
rs975616285 Chr.11: 74706317-74706317 on Build GRCh38
rs763550345 Chr.11: 74706469-74706469 on Build GRCh38
rs558817933 Chr.11: 74706319-74706319 on Build GRCh38
rs745349978 Chr.11: 74706372-74706372 on Build GRCh38
rs774308446 Chr.11: 74706473-74706473 on Build GRCh38
rs112976652 Chr.11: 74706499-74706499 on Build GRCh38
rs11236223 Chr.11: 74706351-74706351 on Build GRCh38
rs371916509 Chr.11: 74706500-74706500 on Build GRCh38
rs993229589 Chr.11: 74706423-74706423 on Build GRCh38
rs767397408 Chr.11: 74706477-74706477 on Build GRCh38
rs758490614 Chr.11: 74706440-74706440 on Build GRCh38
rs139257885 Chr.11: 74706490-74706490 on Build GRCh38
rs906929075 Chr.11: 74706419-74706419 on Build GRCh38
rs527706593 Chr.11: 74706467-74706467 on Build GRCh38
rs1557420 Chr.11: 74706452-74706452 on Build GRCh38
rs747478568 Chr.11: 74706457-74706457 on Build GRCh38
rs71713771 Chr.11: 74706451-74706452 on Build GRCh38
rs778445693 Chr.11: 74706449-74706449 on Build GRCh38
rs555934389 Chr.11: 74706437-74706437 on Build GRCh38
rs184739488 Chr.11: 74706476-74706476 on Build GRCh38
rs987903258 Chr.11: 74706381-74706382 on Build GRCh38
rs561774070 Chr.11: 74706465-74706465 on Build GRCh38
rs541902352 Chr.11: 74706464-74706464 on Build GRCh38
rs377286314 Chr.11: 74706480-74706480 on Build GRCh38
rs779113532 Chr.11: 74706436-74706436 on Build GRCh38
rs200410599 Chr.11: 74706495-74706495 on Build GRCh38
rs1026100325 Chr.11: 74706424-74706424 on Build GRCh38
rs938285279 Chr.11: 74706461-74706461 on Build GRCh38
rs936032034 Chr.11: 74706307-74706307 on Build GRCh38
rs2187044 Chr.11: 74706444-74706444 on Build GRCh38
rs553337811 Chr.11: 74706496-74706496 on Build GRCh38
rs191471502 Chr.11: 74706458-74706458 on Build GRCh38
rs1045981107 Chr.11: 74706391-74706391 on Build GRCh38
rs894419417 Chr.11: 74706355-74706355 on Build GRCh38
rs913683733 Chr.11: 74706411-74706411 on Build GRCh38
rs776836243 Chr.11: 74706462-74706462 on Build GRCh38
rs755868871 Chr.11: 74706489-74706489 on Build GRCh38

View Full Product Details