Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110307844-110308103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A2
Entrez Gene ID
1284
Gene Name
collagen type IV alpha 2
Gene Aliases
ICH, POREN2
UniGene
Hs.508716
Gene Chromosome Location
Chr.13: 110307284-110513027 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5501941 c.45-1G>C p.? Chr.13: 110308068-110308068 on Build GRCh38
1365441 c.41G>A p.R14Q Chr.13: 110307944-110307944 on Build GRCh38
5479788 c.44G>A p.R15Q Chr.13: 110307947-110307947 on Build GRCh38
5823073 c.40C>T p.R14W Chr.13: 110307943-110307943 on Build GRCh38
7183230 c.34G>A p.A12T Chr.13: 110307937-110307937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746438132 Chr.13: 110307972-110307972 on Build GRCh38
rs766265598 Chr.13: 110307947-110307947 on Build GRCh38
rs761596577 Chr.13: 110307930-110307930 on Build GRCh38
rs772772870 Chr.13: 110307986-110307986 on Build GRCh38
rs200970184 Chr.13: 110307909-110307909 on Build GRCh38
rs1025764917 Chr.13: 110307938-110307938 on Build GRCh38
rs746474992 Chr.13: 110307922-110307922 on Build GRCh38
rs963758553 Chr.13: 110307987-110307987 on Build GRCh38
rs569958398 Chr.13: 110308031-110308031 on Build GRCh38
rs768372116 Chr.13: 110307875-110307875 on Build GRCh38
rs773108336 Chr.13: 110307940-110307940 on Build GRCh38
rs754009010 Chr.13: 110308041-110308041 on Build GRCh38
rs369679874 Chr.13: 110307923-110307923 on Build GRCh38
rs759027971 Chr.13: 110308048-110308048 on Build GRCh38
rs754534801 Chr.13: 110307981-110307981 on Build GRCh38
rs561495522 Chr.13: 110307990-110307990 on Build GRCh38
rs913247506 Chr.13: 110308063-110308063 on Build GRCh38
rs766425009 Chr.13: 110307885-110307885 on Build GRCh38
rs200430407 Chr.13: 110308073-110308073 on Build GRCh38
rs781322805 Chr.13: 110307906-110307906 on Build GRCh38
rs374437788 Chr.13: 110307982-110307982 on Build GRCh38
rs751590808 Chr.13: 110307897-110307897 on Build GRCh38
rs552306806 Chr.13: 110308061-110308061 on Build GRCh38
rs752976373 Chr.13: 110307907-110307907 on Build GRCh38
rs770707066 Chr.13: 110308084-110308084 on Build GRCh38
rs760378571 Chr.13: 110307866-110307866 on Build GRCh38
rs776451934 Chr.13: 110307877-110307877 on Build GRCh38
rs147266132 Chr.13: 110308010-110308010 on Build GRCh38
rs755584763 Chr.13: 110308075-110308075 on Build GRCh38
rs759552175 Chr.13: 110307950-110307950 on Build GRCh38
rs767641445 Chr.13: 110307951-110307951 on Build GRCh38
rs777427868 Chr.13: 110308078-110308078 on Build GRCh38
rs769439798 Chr.13: 110307985-110307985 on Build GRCh38
rs769448606 Chr.13: 110307939-110307939 on Build GRCh38
rs779582946 Chr.13: 110307919-110307919 on Build GRCh38
rs747737350 Chr.13: 110307983-110307983 on Build GRCh38
rs759535614 Chr.13: 110308026-110308026 on Build GRCh38
rs34894562 Chr.13: 110307993-110307993 on Build GRCh38
rs762992397 Chr.13: 110307884-110307884 on Build GRCh38
rs376455394 Chr.13: 110307913-110307913 on Build GRCh38
rs763386861 Chr.13: 110307956-110307959 on Build GRCh38
rs1011194662 Chr.13: 110307977-110307977 on Build GRCh38
rs532567032 Chr.13: 110308032-110308032 on Build GRCh38
rs776137096 Chr.13: 110307927-110307927 on Build GRCh38
rs748996226 Chr.13: 110308079-110308079 on Build GRCh38
rs771611881 Chr.13: 110307917-110307917 on Build GRCh38
rs371295752 Chr.13: 110307962-110307962 on Build GRCh38
rs377761467 Chr.13: 110307956-110307956 on Build GRCh38
rs767049610 Chr.13: 110307965-110307973 on Build GRCh38
rs757725450 Chr.13: 110307970-110307970 on Build GRCh38
rs764334376 Chr.13: 110308036-110308036 on Build GRCh38
rs755082963 Chr.13: 110307904-110307904 on Build GRCh38
rs374140752 Chr.13: 110307941-110307941 on Build GRCh38
rs757684961 Chr.13: 110308042-110308042 on Build GRCh38
rs765140800 Chr.13: 110307878-110307878 on Build GRCh38
rs774278201 Chr.13: 110307948-110307948 on Build GRCh38
rs976586162 Chr.13: 110307993-110307993 on Build GRCh38
rs765766348 Chr.13: 110308043-110308043 on Build GRCh38
rs750932950 Chr.13: 110308046-110308046 on Build GRCh38
rs761982058 Chr.13: 110308055-110308055 on Build GRCh38
rs74124101 Chr.13: 110308025-110308025 on Build GRCh38
rs756398337 Chr.13: 110307957-110307957 on Build GRCh38
rs750326918 Chr.13: 110307880-110307880 on Build GRCh38
rs370088110 Chr.13: 110308034-110308034 on Build GRCh38
rs770723628 Chr.13: 110307999-110307999 on Build GRCh38
rs541822832 Chr.13: 110307971-110307971 on Build GRCh38

View Full Product Details