Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34754329-34754594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJD2
Entrez Gene ID
57369
Gene Name
gap junction protein delta 2
Gene Aliases
CX36, GJA9
UniGene
Hs.283816
Gene Chromosome Location
Chr.15: 34751746-34754955 on Build GRCh38
Transcripts
NM_020660.2 XM_017022438.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7130663 c.10T>A p.W4R Chr.15: 34754479-34754479 on Build GRCh38
6671264 c.7delG p.E3fs*9 Chr.15: 34754482-34754482 on Build GRCh38
3711686 c.69G>T p.G23G Chr.15: 34754420-34754420 on Build GRCh38
3500634 c.66C>T p.I22I Chr.15: 34754423-34754423 on Build GRCh38
342832 c.24G>A p.E8E Chr.15: 34754465-34754465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773314897 Chr.15: 34754448-34754448 on Build GRCh38
rs775375152 Chr.15: 34754395-34754395 on Build GRCh38
rs200983881 Chr.15: 34754550-34754550 on Build GRCh38
rs981021833 Chr.15: 34754408-34754408 on Build GRCh38
rs893767438 Chr.15: 34754541-34754541 on Build GRCh38
rs895391554 Chr.15: 34754377-34754377 on Build GRCh38
rs751886847 Chr.15: 34754467-34754467 on Build GRCh38
rs765565710 Chr.15: 34754443-34754443 on Build GRCh38
rs960865184 Chr.15: 34754447-34754447 on Build GRCh38
rs919318458 Chr.15: 34754536-34754536 on Build GRCh38
rs202125808 Chr.15: 34754562-34754562 on Build GRCh38
rs201039869 Chr.15: 34754529-34754529 on Build GRCh38
rs998435636 Chr.15: 34754352-34754352 on Build GRCh38
rs758085315 Chr.15: 34754534-34754534 on Build GRCh38
rs748720896 Chr.15: 34754370-34754370 on Build GRCh38
rs768686463 Chr.15: 34754399-34754399 on Build GRCh38
rs1029376977 Chr.15: 34754364-34754364 on Build GRCh38
rs199874725 Chr.15: 34754544-34754544 on Build GRCh38
rs745444430 Chr.15: 34754387-34754387 on Build GRCh38
rs1009207685 Chr.15: 34754380-34754380 on Build GRCh38
rs763107175 Chr.15: 34754450-34754450 on Build GRCh38
rs746975619 Chr.15: 34754398-34754398 on Build GRCh38
rs984772931 Chr.15: 34754548-34754548 on Build GRCh38
rs373791515 Chr.15: 34754524-34754524 on Build GRCh38
rs200416445 Chr.15: 34754533-34754533 on Build GRCh38
rs763010048 Chr.15: 34754368-34754368 on Build GRCh38
rs369001020 Chr.15: 34754521-34754521 on Build GRCh38
rs761789120 Chr.15: 34754438-34754438 on Build GRCh38
rs200123216 Chr.15: 34754489-34754489 on Build GRCh38
rs187583018 Chr.15: 34754392-34754392 on Build GRCh38
rs766758423 Chr.15: 34754452-34754452 on Build GRCh38
rs370496579 Chr.15: 34754378-34754378 on Build GRCh38
rs776821980 Chr.15: 34754409-34754409 on Build GRCh38
rs767894900 Chr.15: 34754516-34754516 on Build GRCh38
rs535612977 Chr.15: 34754359-34754359 on Build GRCh38
rs200732388 Chr.15: 34754375-34754375 on Build GRCh38
rs77687619 Chr.15: 34754384-34754384 on Build GRCh38
rs201869332 Chr.15: 34754543-34754543 on Build GRCh38

View Full Product Details