Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157162339-157162600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM71B
Entrez Gene ID
153745
Gene Name
family with sequence similarity 71 member B
Gene Aliases
-
UniGene
Hs.666099
Gene Chromosome Location
Chr.5: 157162332-157166268 on Build GRCh38
Transcripts
NM_130899.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
304015 c.1729G>A p.A577T Chr.5: 157162536-157162536 on Build GRCh38
3975360 c.1672G>A p.V558I Chr.5: 157162593-157162593 on Build GRCh38
106361 c.1794C>T p.I598I Chr.5: 157162471-157162471 on Build GRCh38
6353322 c.1795G>A p.V599I Chr.5: 157162470-157162470 on Build GRCh38
5996183 c.1800T>C p.F600F Chr.5: 157162465-157162465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144807004 Chr.5: 157162480-157162480 on Build GRCh38
rs550645272 Chr.5: 157162427-157162427 on Build GRCh38
rs943356286 Chr.5: 157162393-157162393 on Build GRCh38
rs770794585 Chr.5: 157162481-157162481 on Build GRCh38
rs779120509 Chr.5: 157162431-157162431 on Build GRCh38
rs907699503 Chr.5: 157162379-157162379 on Build GRCh38
rs758740726 Chr.5: 157162504-157162504 on Build GRCh38
rs892845541 Chr.5: 157162493-157162493 on Build GRCh38
rs2278385 Chr.5: 157162388-157162388 on Build GRCh38
rs761214256 Chr.5: 157162423-157162423 on Build GRCh38
rs31208 Chr.5: 157162574-157162574 on Build GRCh38
rs149402782 Chr.5: 157162455-157162455 on Build GRCh38
rs753288128 Chr.5: 157162412-157162412 on Build GRCh38
rs764423052 Chr.5: 157162425-157162425 on Build GRCh38
rs754238315 Chr.5: 157162519-157162519 on Build GRCh38
rs1039423415 Chr.5: 157162394-157162394 on Build GRCh38
rs370147732 Chr.5: 157162395-157162395 on Build GRCh38
rs1011737977 Chr.5: 157162518-157162518 on Build GRCh38
rs751658693 Chr.5: 157162559-157162559 on Build GRCh38
rs1001806042 Chr.5: 157162515-157162515 on Build GRCh38
rs1047845304 Chr.5: 157162443-157162443 on Build GRCh38
rs775118965 Chr.5: 157162489-157162489 on Build GRCh38
rs199813925 Chr.5: 157162508-157162508 on Build GRCh38
rs759132386 Chr.5: 157162403-157162403 on Build GRCh38
rs758593330 Chr.5: 157162546-157162546 on Build GRCh38
rs899180691 Chr.5: 157162402-157162402 on Build GRCh38
rs531731172 Chr.5: 157162424-157162424 on Build GRCh38
rs765543128 Chr.5: 157162537-157162537 on Build GRCh38
rs995298335 Chr.5: 157162439-157162439 on Build GRCh38
rs374704685 Chr.5: 157162482-157162482 on Build GRCh38
rs2115480 Chr.5: 157162470-157162470 on Build GRCh38
rs769761529 Chr.5: 157162479-157162479 on Build GRCh38
rs780169350 Chr.5: 157162553-157162553 on Build GRCh38
rs767088416 Chr.5: 157162407-157162407 on Build GRCh38
rs147391845 Chr.5: 157162471-157162471 on Build GRCh38
rs750679969 Chr.5: 157162542-157162542 on Build GRCh38
rs574910592 Chr.5: 157162377-157162377 on Build GRCh38
rs759225645 Chr.5: 157162483-157162483 on Build GRCh38
rs1051624189 Chr.5: 157162372-157162372 on Build GRCh38
rs762258472 Chr.5: 157162531-157162531 on Build GRCh38
rs369804995 Chr.5: 157162434-157162434 on Build GRCh38
rs148633922 Chr.5: 157162488-157162488 on Build GRCh38
rs750592442 Chr.5: 157162433-157162433 on Build GRCh38
rs754225436 Chr.5: 157162426-157162426 on Build GRCh38

View Full Product Details