Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 59182299-59182524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTX4
Entrez Gene ID
23220
Gene Name
deltex E3 ubiquitin ligase 4
Gene Aliases
RNF155
UniGene
Hs.523696
Gene Chromosome Location
Chr.11: 59171430-59208587 on Build GRCh38
Transcripts
NM_001300727.1 NM_015177.1 XM_006718483.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5838879 c.848A>G p.K283R Chr.11: 59182375-59182375 on Build GRCh38
6223181 c.865C>A p.L289M Chr.11: 59182392-59182392 on Build GRCh38
4679631 c.775C>T p.R259W Chr.11: 59182302-59182302 on Build GRCh38
6223182 c.547C>A p.L183M Chr.11: 59182392-59182392 on Build GRCh38
5896113 c.880C>T p.R294C Chr.11: 59182407-59182407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772953218 Chr.11: 59182398-59182398 on Build GRCh38
rs772601790 Chr.11: 59182374-59182374 on Build GRCh38
rs942934596 Chr.11: 59182362-59182362 on Build GRCh38
rs766052306 Chr.11: 59182353-59182353 on Build GRCh38
rs370051674 Chr.11: 59182333-59182333 on Build GRCh38
rs754303075 Chr.11: 59182363-59182363 on Build GRCh38
rs759176432 Chr.11: 59182482-59182482 on Build GRCh38
rs372858846 Chr.11: 59182443-59182443 on Build GRCh38
rs769903609 Chr.11: 59182449-59182449 on Build GRCh38
rs780353069 Chr.11: 59182433-59182433 on Build GRCh38
rs750233515 Chr.11: 59182348-59182348 on Build GRCh38
rs770643875 Chr.11: 59182468-59182468 on Build GRCh38
rs764132824 Chr.11: 59182337-59182337 on Build GRCh38
rs757795127 Chr.11: 59182412-59182412 on Build GRCh38
rs760781760 Chr.11: 59182336-59182336 on Build GRCh38
rs774464332 Chr.11: 59182396-59182396 on Build GRCh38
rs779369409 Chr.11: 59182370-59182370 on Build GRCh38
rs761753873 Chr.11: 59182390-59182390 on Build GRCh38
rs776581480 Chr.11: 59182386-59182386 on Build GRCh38
rs374017839 Chr.11: 59182339-59182339 on Build GRCh38
rs747191229 Chr.11: 59182435-59182435 on Build GRCh38
rs1032361380 Chr.11: 59182421-59182421 on Build GRCh38
rs758826918 Chr.11: 59182429-59182429 on Build GRCh38
rs775244664 Chr.11: 59182489-59182489 on Build GRCh38
rs780405637 Chr.11: 59182378-59182378 on Build GRCh38
rs1035568163 Chr.11: 59182437-59182437 on Build GRCh38
rs375108646 Chr.11: 59182401-59182401 on Build GRCh38
rs141860745 Chr.11: 59182408-59182408 on Build GRCh38
rs199765629 Chr.11: 59182455-59182455 on Build GRCh38
rs766220329 Chr.11: 59182407-59182407 on Build GRCh38
rs748258535 Chr.11: 59182446-59182446 on Build GRCh38
rs1047209012 Chr.11: 59182400-59182400 on Build GRCh38
rs930122597 Chr.11: 59182397-59182397 on Build GRCh38
rs370919698 Chr.11: 59182380-59182380 on Build GRCh38
rs368757950 Chr.11: 59182423-59182423 on Build GRCh38
rs571331933 Chr.11: 59182331-59182331 on Build GRCh38
rs764946673 Chr.11: 59182346-59182346 on Build GRCh38
rs747888008 Chr.11: 59182411-59182411 on Build GRCh38
rs200340632 Chr.11: 59182444-59182444 on Build GRCh38
rs762544733 Chr.11: 59182351-59182351 on Build GRCh38
rs75403375 Chr.11: 59182379-59182379 on Build GRCh38
rs774028305 Chr.11: 59182480-59182480 on Build GRCh38
rs199758601 Chr.11: 59182359-59182359 on Build GRCh38
rs375750062 Chr.11: 59182325-59182325 on Build GRCh38
rs767105404 Chr.11: 59182485-59182485 on Build GRCh38
rs761725969 Chr.11: 59182343-59182343 on Build GRCh38
rs533589791 Chr.11: 59182366-59182366 on Build GRCh38
rs762944983 Chr.11: 59182467-59182467 on Build GRCh38
rs774857541 Chr.11: 59182327-59182327 on Build GRCh38
rs751219014 Chr.11: 59182358-59182358 on Build GRCh38
rs373646554 Chr.11: 59182361-59182361 on Build GRCh38

View Full Product Details