Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123626977-123627250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B1
Entrez Gene ID
1967
Gene Name
eukaryotic translation initiation factor 2B subunit alpha
Gene Aliases
EIF2B, EIF2BA
UniGene
Hs.741273
Gene Chromosome Location
Chr.12: 123621023-123633776 on Build GRCh38
Transcripts
NM_001414.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4039779 c.449T>C p.V150A Chr.12: 123627077-123627077 on Build GRCh38
4493876 c.422C>T p.A141V Chr.12: 123627104-123627104 on Build GRCh38
5840442 c.430G>A p.A144T Chr.12: 123627096-123627096 on Build GRCh38
3457538 c.411C>T p.V137V Chr.12: 123627115-123627115 on Build GRCh38
4825729 c.381T>A p.T127T Chr.12: 123627145-123627145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774774980 Chr.12: 123627005-123627005 on Build GRCh38
rs759473946 Chr.12: 123627195-123627195 on Build GRCh38
rs574891460 Chr.12: 123627154-123627154 on Build GRCh38
rs778317602 Chr.12: 123627153-123627153 on Build GRCh38
rs752937669 Chr.12: 123627162-123627162 on Build GRCh38
rs1044972101 Chr.12: 123627223-123627223 on Build GRCh38
rs758934938 Chr.12: 123627032-123627032 on Build GRCh38
rs7298287 Chr.12: 123627176-123627176 on Build GRCh38
rs762724593 Chr.12: 123627042-123627042 on Build GRCh38
rs1022900838 Chr.12: 123627029-123627029 on Build GRCh38
rs760625796 Chr.12: 123627191-123627191 on Build GRCh38
rs760720371 Chr.12: 123627114-123627114 on Build GRCh38
rs146748240 Chr.12: 123627142-123627142 on Build GRCh38
rs748469050 Chr.12: 123627078-123627078 on Build GRCh38
rs769237783 Chr.12: 123627156-123627156 on Build GRCh38
rs748630164 Chr.12: 123627007-123627007 on Build GRCh38
rs888740026 Chr.12: 123627040-123627040 on Build GRCh38
rs750323816 Chr.12: 123627112-123627112 on Build GRCh38
rs974189720 Chr.12: 123627170-123627170 on Build GRCh38
rs377035152 Chr.12: 123627179-123627179 on Build GRCh38
rs896345569 Chr.12: 123627216-123627216 on Build GRCh38
rs776148289 Chr.12: 123627017-123627017 on Build GRCh38
rs73414297 Chr.12: 123627109-123627109 on Build GRCh38
rs148714712 Chr.12: 123627097-123627097 on Build GRCh38
rs773328292 Chr.12: 123627183-123627183 on Build GRCh38
rs1001293045 Chr.12: 123627150-123627150 on Build GRCh38
rs377608966 Chr.12: 123627080-123627080 on Build GRCh38
rs751503002 Chr.12: 123627049-123627049 on Build GRCh38
rs750555015 Chr.12: 123627064-123627064 on Build GRCh38
rs755256108 Chr.12: 123627141-123627141 on Build GRCh38
rs766456869 Chr.12: 123627133-123627133 on Build GRCh38
rs767525555 Chr.12: 123627058-123627058 on Build GRCh38
rs774320069 Chr.12: 123627098-123627098 on Build GRCh38
rs780061310 Chr.12: 123627142-123627143 on Build GRCh38
rs775140578 Chr.12: 123627041-123627041 on Build GRCh38
rs951410731 Chr.12: 123627229-123627229 on Build GRCh38
rs749631884 Chr.12: 123626999-123626999 on Build GRCh38
rs779302767 Chr.12: 123627077-123627077 on Build GRCh38
rs200645851 Chr.12: 123627046-123627046 on Build GRCh38
rs766184154 Chr.12: 123627194-123627194 on Build GRCh38
rs745385760 Chr.12: 123627086-123627086 on Build GRCh38
rs202056385 Chr.12: 123627158-123627158 on Build GRCh38
rs768380909 Chr.12: 123627166-123627166 on Build GRCh38
rs765448419 Chr.12: 123627198-123627198 on Build GRCh38
rs756209363 Chr.12: 123627070-123627070 on Build GRCh38
rs770465765 Chr.12: 123627016-123627016 on Build GRCh38
rs780147875 Chr.12: 123627072-123627072 on Build GRCh38
rs1033102236 Chr.12: 123627079-123627079 on Build GRCh38
rs139913280 Chr.12: 123627102-123627102 on Build GRCh38
rs947948195 Chr.12: 123627222-123627222 on Build GRCh38
rs774092291 Chr.12: 123627175-123627175 on Build GRCh38
rs149510594 Chr.12: 123627076-123627076 on Build GRCh38
rs769344308 Chr.12: 123627037-123627037 on Build GRCh38
rs754044114 Chr.12: 123627137-123627137 on Build GRCh38
rs758695418 Chr.12: 123627152-123627152 on Build GRCh38
rs369712292 Chr.12: 123627002-123627002 on Build GRCh38
rs190571435 Chr.12: 123627224-123627224 on Build GRCh38
rs199625718 Chr.12: 123627061-123627061 on Build GRCh38
rs549587142 Chr.12: 123627073-123627073 on Build GRCh38
rs748708533 Chr.12: 123627165-123627165 on Build GRCh38
rs370678173 Chr.12: 123627087-123627087 on Build GRCh38
rs753071106 Chr.12: 123627145-123627145 on Build GRCh38
rs752911399 Chr.12: 123627204-123627204 on Build GRCh38
rs758995973 Chr.12: 123627020-123627020 on Build GRCh38
rs193142170 Chr.12: 123627051-123627051 on Build GRCh38
rs762644737 Chr.12: 123627096-123627096 on Build GRCh38

View Full Product Details