Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35642636-35642875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV2
Entrez Gene ID
2116
Gene Name
ETS variant 2
Gene Aliases
ER71, ETSRP71
UniGene
Hs.194061
Gene Chromosome Location
Chr.19: 35641175-35644871 on Build GRCh38
Transcripts
NM_001300974.1 NM_001304549.1 NM_014209.3 XM_005258652.2 XM_011526624.2 XM_017026472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6999117 c.122C>T p.T41M Chr.19: 35642666-35642666 on Build GRCh38
6439965 c.117_118GG>AA p.D40N Chr.19: 35642661-35642662 on Build GRCh38
1205747 c.215C>T p.A72V Chr.19: 35642675-35642675 on Build GRCh38
6999116 c.206C>T p.T69M Chr.19: 35642666-35642666 on Build GRCh38
6439964 c.201_202GG>AA p.D68N Chr.19: 35642661-35642662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924379840 Chr.19: 35642770-35642771 on Build GRCh38
rs546013638 Chr.19: 35642664-35642664 on Build GRCh38
rs533941122 Chr.19: 35642851-35642851 on Build GRCh38
rs1022712637 Chr.19: 35642759-35642759 on Build GRCh38
rs934431614 Chr.19: 35642812-35642812 on Build GRCh38
rs568485038 Chr.19: 35642847-35642847 on Build GRCh38
rs757341669 Chr.19: 35642804-35642804 on Build GRCh38
rs763630007 Chr.19: 35642715-35642716 on Build GRCh38
rs751755067 Chr.19: 35642769-35642769 on Build GRCh38
rs764848652 Chr.19: 35642714-35642714 on Build GRCh38
rs971261410 Chr.19: 35642765-35642765 on Build GRCh38
rs752245434 Chr.19: 35642735-35642735 on Build GRCh38
rs57526070 Chr.19: 35642844-35642844 on Build GRCh38
rs776976052 Chr.19: 35642678-35642678 on Build GRCh38
rs989803902 Chr.19: 35642817-35642817 on Build GRCh38
rs144701792 Chr.19: 35642742-35642742 on Build GRCh38
rs190349714 Chr.19: 35642675-35642675 on Build GRCh38
rs757959930 Chr.19: 35642738-35642738 on Build GRCh38
rs912296490 Chr.19: 35642657-35642657 on Build GRCh38
rs759245318 Chr.19: 35642707-35642707 on Build GRCh38
rs951934747 Chr.19: 35642739-35642739 on Build GRCh38

View Full Product Details