Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43306322-43306547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIP3
Entrez Gene ID
401262
Gene Name
cysteine rich protein 3
Gene Aliases
TLP, TLP-A, bA480N24.2
UniGene
Hs.653165
Gene Chromosome Location
Chr.6: 43305473-43310705 on Build GRCh38
Transcripts
NM_206922.2 XM_005249103.3 XM_011514609.2 XM_017010856.1 XM_017010857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3627616 c.365C>T p.S122L Chr.6: 43306481-43306481 on Build GRCh38
4925576 c.381T>C p.C127C Chr.6: 43306465-43306465 on Build GRCh38
5820139 c.395A>C p.Y132S Chr.6: 43306451-43306451 on Build GRCh38
4922751 c.391G>A p.V131I Chr.6: 43306455-43306455 on Build GRCh38
1079544 c.366G>A p.S122S Chr.6: 43306480-43306480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890781810 Chr.6: 43306442-43306442 on Build GRCh38
rs972607443 Chr.6: 43306378-43306378 on Build GRCh38
rs774622516 Chr.6: 43306432-43306432 on Build GRCh38
rs760463930 Chr.6: 43306456-43306456 on Build GRCh38
rs761139953 Chr.6: 43306438-43306441 on Build GRCh38
rs750395224 Chr.6: 43306455-43306455 on Build GRCh38
rs761830344 Chr.6: 43306451-43306451 on Build GRCh38
rs777653358 Chr.6: 43306480-43306480 on Build GRCh38
rs746372397 Chr.6: 43306508-43306508 on Build GRCh38
rs373267999 Chr.6: 43306452-43306452 on Build GRCh38
rs753487236 Chr.6: 43306470-43306470 on Build GRCh38
rs899060993 Chr.6: 43306385-43306385 on Build GRCh38
rs774738343 Chr.6: 43306420-43306420 on Build GRCh38
rs766112718 Chr.6: 43306466-43306466 on Build GRCh38
rs770057505 Chr.6: 43306398-43306398 on Build GRCh38
rs757755897 Chr.6: 43306493-43306493 on Build GRCh38
rs369154404 Chr.6: 43306421-43306421 on Build GRCh38
rs371829804 Chr.6: 43306410-43306410 on Build GRCh38
rs751322556 Chr.6: 43306518-43306518 on Build GRCh38
rs752259267 Chr.6: 43306479-43306479 on Build GRCh38
rs768751193 Chr.6: 43306429-43306429 on Build GRCh38
rs376130095 Chr.6: 43306412-43306412 on Build GRCh38
rs1019117990 Chr.6: 43306488-43306488 on Build GRCh38
rs879713441 Chr.6: 43306483-43306483 on Build GRCh38
rs918345636 Chr.6: 43306386-43306386 on Build GRCh38
rs530236387 Chr.6: 43306528-43306528 on Build GRCh38
rs756451115 Chr.6: 43306517-43306517 on Build GRCh38
rs754624385 Chr.6: 43306473-43306473 on Build GRCh38
rs781612773 Chr.6: 43306504-43306504 on Build GRCh38
rs746309926 Chr.6: 43306395-43306395 on Build GRCh38
rs771473394 Chr.6: 43306359-43306359 on Build GRCh38
rs764700704 Chr.6: 43306475-43306475 on Build GRCh38

View Full Product Details