Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5603327-5603574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSCN1
Entrez Gene ID
6624
Gene Name
fascin actin-bundling protein 1
Gene Aliases
FAN1, HSN, SNL, p55
UniGene
Hs.118400
Gene Chromosome Location
Chr.7: 5592805-5606656 on Build GRCh38
Transcripts
NM_003088.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5997470 c.1026C>G p.D342E Chr.7: 5603532-5603532 on Build GRCh38
6036131 c.1021C>T p.R341C Chr.7: 5603527-5603527 on Build GRCh38
7234595 c.1008T>C p.F336F Chr.7: 5603514-5603514 on Build GRCh38
7271680 c.1014C>T p.I338I Chr.7: 5603520-5603520 on Build GRCh38
7368867 c.932C>T p.T311M Chr.7: 5603356-5603356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760536657 Chr.7: 5603378-5603378 on Build GRCh38
rs559133408 Chr.7: 5603468-5603468 on Build GRCh38
rs193001737 Chr.7: 5603420-5603420 on Build GRCh38
rs751959319 Chr.7: 5603391-5603391 on Build GRCh38
rs374138413 Chr.7: 5603441-5603441 on Build GRCh38
rs143738037 Chr.7: 5603371-5603371 on Build GRCh38
rs1640233 Chr.7: 5603514-5603514 on Build GRCh38
rs199969674 Chr.7: 5603442-5603442 on Build GRCh38
rs748836930 Chr.7: 5603424-5603424 on Build GRCh38
rs372102668 Chr.7: 5603469-5603469 on Build GRCh38
rs766746574 Chr.7: 5603550-5603550 on Build GRCh38
rs757174383 Chr.7: 5603347-5603347 on Build GRCh38
rs758659055 Chr.7: 5603389-5603389 on Build GRCh38
rs763256193 Chr.7: 5603449-5603450 on Build GRCh38
rs778403057 Chr.7: 5603483-5603483 on Build GRCh38
rs776651945 Chr.7: 5603515-5603515 on Build GRCh38
rs150576558 Chr.7: 5603377-5603377 on Build GRCh38
rs754431173 Chr.7: 5603471-5603471 on Build GRCh38
rs201921858 Chr.7: 5603478-5603478 on Build GRCh38
rs770227239 Chr.7: 5603407-5603407 on Build GRCh38
rs561836811 Chr.7: 5603428-5603428 on Build GRCh38
rs757449828 Chr.7: 5603452-5603452 on Build GRCh38
rs11557822 Chr.7: 5603384-5603384 on Build GRCh38
rs201909136 Chr.7: 5603440-5603440 on Build GRCh38
rs773024617 Chr.7: 5603489-5603489 on Build GRCh38
rs573612458 Chr.7: 5603438-5603438 on Build GRCh38
rs369111423 Chr.7: 5603487-5603487 on Build GRCh38
rs961286728 Chr.7: 5603492-5603492 on Build GRCh38
rs767673781 Chr.7: 5603534-5603534 on Build GRCh38
rs772902047 Chr.7: 5603437-5603437 on Build GRCh38
rs771836243 Chr.7: 5603436-5603436 on Build GRCh38
rs774051314 Chr.7: 5603426-5603426 on Build GRCh38
rs186765948 Chr.7: 5603536-5603536 on Build GRCh38
rs762074681 Chr.7: 5603533-5603533 on Build GRCh38
rs201344792 Chr.7: 5603484-5603484 on Build GRCh38
rs111761155 Chr.7: 5603356-5603356 on Build GRCh38
rs748787713 Chr.7: 5603470-5603470 on Build GRCh38
rs73052796 Chr.7: 5603348-5603348 on Build GRCh38
rs764227580 Chr.7: 5603521-5603521 on Build GRCh38
rs774102065 Chr.7: 5603363-5603363 on Build GRCh38
rs767346804 Chr.7: 5603372-5603372 on Build GRCh38
rs748357218 Chr.7: 5603527-5603527 on Build GRCh38
rs184652662 Chr.7: 5603493-5603493 on Build GRCh38
rs200736047 Chr.7: 5603425-5603425 on Build GRCh38
rs778208354 Chr.7: 5603390-5603390 on Build GRCh38
rs781573524 Chr.7: 5603466-5603466 on Build GRCh38
rs751736271 Chr.7: 5603446-5603446 on Build GRCh38
rs757583431 Chr.7: 5603397-5603397 on Build GRCh38
rs756533788 Chr.7: 5603537-5603537 on Build GRCh38
rs377275041 Chr.7: 5603528-5603528 on Build GRCh38
rs754279900 Chr.7: 5603552-5603552 on Build GRCh38
rs374500646 Chr.7: 5603402-5603402 on Build GRCh38
rs752869249 Chr.7: 5603387-5603387 on Build GRCh38

View Full Product Details