Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100678043-100678302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB2
Entrez Gene ID
2783
Gene Name
G protein subunit beta 2
Gene Aliases
-
UniGene
Hs.185172
Gene Chromosome Location
Chr.7: 100673740-100679169 on Build GRCh38
Transcripts
NM_005273.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208409 c.682G>A p.D228N Chr.7: 100678282-100678282 on Build GRCh38
1292385 c.571T>G p.S191A Chr.7: 100678171-100678171 on Build GRCh38
4428584 c.580C>G p.P194A Chr.7: 100678180-100678180 on Build GRCh38
6444504 c.671_672GC>TT p.G224V Chr.7: 100678271-100678272 on Build GRCh38
1673378 c.535G>T p.G179C Chr.7: 100678135-100678135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923325507 Chr.7: 100678224-100678224 on Build GRCh38
rs769501179 Chr.7: 100678177-100678177 on Build GRCh38
rs371447650 Chr.7: 100678269-100678269 on Build GRCh38
rs139214321 Chr.7: 100678190-100678190 on Build GRCh38
rs770474986 Chr.7: 100678064-100678068 on Build GRCh38
rs149502189 Chr.7: 100678242-100678242 on Build GRCh38
rs200586443 Chr.7: 100678094-100678094 on Build GRCh38
rs574237260 Chr.7: 100678239-100678239 on Build GRCh38
rs781496683 Chr.7: 100678234-100678234 on Build GRCh38
rs2406154 Chr.7: 100678146-100678146 on Build GRCh38
rs368070921 Chr.7: 100678194-100678194 on Build GRCh38
rs781614369 Chr.7: 100678171-100678171 on Build GRCh38
rs762496734 Chr.7: 100678069-100678069 on Build GRCh38
rs1016962982 Chr.7: 100678101-100678101 on Build GRCh38
rs756394669 Chr.7: 100678237-100678237 on Build GRCh38
rs373871017 Chr.7: 100678182-100678182 on Build GRCh38
rs542867104 Chr.7: 100678226-100678226 on Build GRCh38
rs755355032 Chr.7: 100678161-100678161 on Build GRCh38
rs766442414 Chr.7: 100678070-100678070 on Build GRCh38
rs753129673 Chr.7: 100678236-100678236 on Build GRCh38
rs141859943 Chr.7: 100678179-100678179 on Build GRCh38
rs1050520443 Chr.7: 100678267-100678267 on Build GRCh38
rs748396013 Chr.7: 100678176-100678176 on Build GRCh38
rs374877478 Chr.7: 100678275-100678275 on Build GRCh38
rs770626958 Chr.7: 100678281-100678281 on Build GRCh38
rs780578984 Chr.7: 100678134-100678134 on Build GRCh38
rs769947840 Chr.7: 100678067-100678068 on Build GRCh38
rs889272812 Chr.7: 100678272-100678272 on Build GRCh38
rs766432973 Chr.7: 100678220-100678220 on Build GRCh38
rs374626655 Chr.7: 100678063-100678063 on Build GRCh38
rs774483254 Chr.7: 100678189-100678189 on Build GRCh38
rs754068566 Chr.7: 100678100-100678100 on Build GRCh38
rs752126417 Chr.7: 100678222-100678222 on Build GRCh38
rs983636308 Chr.7: 100678090-100678090 on Build GRCh38
rs747491823 Chr.7: 100678149-100678149 on Build GRCh38
rs79631023 Chr.7: 100678183-100678183 on Build GRCh38
rs758428726 Chr.7: 100678132-100678132 on Build GRCh38
rs150959922 Chr.7: 100678131-100678131 on Build GRCh38
rs200210649 Chr.7: 100678206-100678206 on Build GRCh38
rs5001693 Chr.7: 100678209-100678209 on Build GRCh38
rs138214277 Chr.7: 100678120-100678120 on Build GRCh38
rs771831778 Chr.7: 100678193-100678193 on Build GRCh38
rs757499352 Chr.7: 100678119-100678119 on Build GRCh38
rs145101383 Chr.7: 100678089-100678089 on Build GRCh38
rs376741553 Chr.7: 100678083-100678083 on Build GRCh38

View Full Product Details