Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 65809597-65809858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH7
Entrez Gene ID
1005
Gene Name
cadherin 7
Gene Aliases
CDH7L1
UniGene
Hs.541003
Gene Chromosome Location
Chr.18: 65750252-65885136 on Build GRCh38
Transcripts
NM_001317214.1 NM_004361.3 NM_033646.2 XM_017025523.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5921313 c.275G>T p.S92I Chr.18: 65809768-65809768 on Build GRCh38
391126 c.342delG p.E114fs*62 Chr.18: 65809835-65809835 on Build GRCh38
1730895 c.227A>G p.D76G Chr.18: 65809720-65809720 on Build GRCh38
1480427 c.233G>C p.G78A Chr.18: 65809726-65809726 on Build GRCh38
4951056 c.238G>T p.G80C Chr.18: 65809731-65809731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910886235 Chr.18: 65809744-65809744 on Build GRCh38
rs746015569 Chr.18: 65809820-65809820 on Build GRCh38
rs749985277 Chr.18: 65809749-65809749 on Build GRCh38
rs778300269 Chr.18: 65809785-65809785 on Build GRCh38
rs1033507611 Chr.18: 65809670-65809670 on Build GRCh38
rs563642396 Chr.18: 65809835-65809835 on Build GRCh38
rs371286997 Chr.18: 65809732-65809732 on Build GRCh38
rs779910675 Chr.18: 65809758-65809758 on Build GRCh38
rs1014154451 Chr.18: 65809627-65809627 on Build GRCh38
rs748465617 Chr.18: 65809768-65809768 on Build GRCh38
rs545060438 Chr.18: 65809651-65809651 on Build GRCh38
rs773605077 Chr.18: 65809655-65809655 on Build GRCh38
rs747494494 Chr.18: 65809796-65809796 on Build GRCh38
rs934262499 Chr.18: 65809751-65809751 on Build GRCh38
rs1024155365 Chr.18: 65809628-65809628 on Build GRCh38
rs61741837 Chr.18: 65809757-65809757 on Build GRCh38
rs762797509 Chr.18: 65809715-65809715 on Build GRCh38
rs913196076 Chr.18: 65809750-65809750 on Build GRCh38
rs955136254 Chr.18: 65809668-65809668 on Build GRCh38
rs763964939 Chr.18: 65809723-65809723 on Build GRCh38
rs114235038 Chr.18: 65809746-65809746 on Build GRCh38
rs770944914 Chr.18: 65809659-65809659 on Build GRCh38
rs756740303 Chr.18: 65809745-65809745 on Build GRCh38
rs774247887 Chr.18: 65809839-65809839 on Build GRCh38
rs776542846 Chr.18: 65809800-65809800 on Build GRCh38
rs867768827 Chr.18: 65809726-65809726 on Build GRCh38
rs765483899 Chr.18: 65809678-65809678 on Build GRCh38
rs758837028 Chr.18: 65809773-65809773 on Build GRCh38
rs751197051 Chr.18: 65809810-65809810 on Build GRCh38
rs769993400 Chr.18: 65809824-65809824 on Build GRCh38
rs776866961 Chr.18: 65809672-65809672 on Build GRCh38
rs957426531 Chr.18: 65809730-65809730 on Build GRCh38
rs942845414 Chr.18: 65809840-65809840 on Build GRCh38
rs763699847 Chr.18: 65809661-65809662 on Build GRCh38
rs149963244 Chr.18: 65809653-65809653 on Build GRCh38
rs760740890 Chr.18: 65809656-65809656 on Build GRCh38
rs775706781 Chr.18: 65809827-65809827 on Build GRCh38
rs751458351 Chr.18: 65809738-65809738 on Build GRCh38
rs759733898 Chr.18: 65809675-65809675 on Build GRCh38
rs374844505 Chr.18: 65809660-65809660 on Build GRCh38
rs369254854 Chr.18: 65809707-65809707 on Build GRCh38
rs868447230 Chr.18: 65809828-65809828 on Build GRCh38

View Full Product Details