Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35893052-35893323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGO1
Entrez Gene ID
26523
Gene Name
argonaute 1, RISC catalytic component
Gene Aliases
EIF2C, EIF2C1, GERP95, Q99, hAgo1
UniGene
Hs.22867
Gene Chromosome Location
Chr.1: 35869808-35929610 on Build GRCh38
Transcripts
NM_001317122.1 NM_001317123.1 NM_012199.4 XM_011541236.2 XM_017001016.1 XM_017001017.1 XM_017001018.1 XM_017001019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6507181 c.340G>A p.E114K Chr.1: 35893106-35893106 on Build GRCh38
680927 c.342G>T p.E114D Chr.1: 35893108-35893108 on Build GRCh38
6323316 c.395G>T p.W132L Chr.1: 35893161-35893161 on Build GRCh38
4007414 c.491T>C p.M164T Chr.1: 35893257-35893257 on Build GRCh38
3733636 c.347C>A p.T116K Chr.1: 35893113-35893113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374386582 Chr.1: 35893074-35893074 on Build GRCh38
rs768140024 Chr.1: 35893159-35893159 on Build GRCh38
rs774463509 Chr.1: 35893114-35893114 on Build GRCh38
rs772403603 Chr.1: 35893273-35893273 on Build GRCh38
rs778531571 Chr.1: 35893201-35893201 on Build GRCh38
rs774413049 Chr.1: 35893239-35893239 on Build GRCh38
rs771183369 Chr.1: 35893237-35893237 on Build GRCh38
rs200107860 Chr.1: 35893268-35893268 on Build GRCh38
rs140048484 Chr.1: 35893198-35893198 on Build GRCh38
rs763796047 Chr.1: 35893186-35893186 on Build GRCh38
rs780052272 Chr.1: 35893208-35893208 on Build GRCh38
rs375926535 Chr.1: 35893085-35893085 on Build GRCh38
rs1000221753 Chr.1: 35893252-35893252 on Build GRCh38
rs750019410 Chr.1: 35893204-35893204 on Build GRCh38
rs766471361 Chr.1: 35893100-35893100 on Build GRCh38
rs867858441 Chr.1: 35893101-35893101 on Build GRCh38
rs551846586 Chr.1: 35893207-35893207 on Build GRCh38
rs974811600 Chr.1: 35893169-35893169 on Build GRCh38
rs773426159 Chr.1: 35893082-35893082 on Build GRCh38
rs762981843 Chr.1: 35893090-35893090 on Build GRCh38
rs146336392 Chr.1: 35893231-35893231 on Build GRCh38
rs372148672 Chr.1: 35893291-35893291 on Build GRCh38
rs777924524 Chr.1: 35893225-35893225 on Build GRCh38
rs759723959 Chr.1: 35893256-35893256 on Build GRCh38
rs756518480 Chr.1: 35893185-35893185 on Build GRCh38
rs748561732 Chr.1: 35893182-35893182 on Build GRCh38
rs768528277 Chr.1: 35893224-35893224 on Build GRCh38
rs746713624 Chr.1: 35893217-35893217 on Build GRCh38
rs759929800 Chr.1: 35893144-35893144 on Build GRCh38
rs769848231 Chr.1: 35893075-35893075 on Build GRCh38
rs1044509129 Chr.1: 35893214-35893214 on Build GRCh38
rs368040201 Chr.1: 35893078-35893078 on Build GRCh38
rs1037229267 Chr.1: 35893099-35893099 on Build GRCh38

View Full Product Details