Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 172664211-172664456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASLG
Entrez Gene ID
356
Gene Name
Fas ligand
Gene Aliases
ALPS1B, APT1LG1, APTL, CD178, CD95-L, CD95L, FASL, TNFSF6, TNLG1A
UniGene
Hs.2007
Gene Chromosome Location
Chr.1: 172659008-172666873 on Build GRCh38
Transcripts
NM_000639.2 NM_001302746.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4547845 c.432G>A p.R144R Chr.1: 172664371-172664371 on Build GRCh38
3705245 c.413C>A p.P138H Chr.1: 172664352-172664352 on Build GRCh38
6357124 c.404G>T p.S135I Chr.1: 172664343-172664343 on Build GRCh38
6315503 c.451+5G>T p.? Chr.1: 172664395-172664395 on Build GRCh38
4933045 c.402C>T p.P134P Chr.1: 172664341-172664341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049754208 Chr.1: 172664338-172664338 on Build GRCh38
rs767244050 Chr.1: 172664259-172664259 on Build GRCh38
rs763580780 Chr.1: 172664425-172664425 on Build GRCh38
rs945505342 Chr.1: 172664243-172664244 on Build GRCh38
rs143306673 Chr.1: 172664341-172664341 on Build GRCh38
rs777090474 Chr.1: 172664414-172664414 on Build GRCh38
rs750816051 Chr.1: 172664320-172664320 on Build GRCh38
rs971837868 Chr.1: 172664367-172664367 on Build GRCh38
rs368143171 Chr.1: 172664355-172664355 on Build GRCh38
rs913033399 Chr.1: 172664252-172664252 on Build GRCh38
rs1042939495 Chr.1: 172664277-172664277 on Build GRCh38
rs893885885 Chr.1: 172664242-172664242 on Build GRCh38
rs752291238 Chr.1: 172664409-172664409 on Build GRCh38
rs780093284 Chr.1: 172664356-172664356 on Build GRCh38
rs745583712 Chr.1: 172664354-172664354 on Build GRCh38
rs756917304 Chr.1: 172664351-172664351 on Build GRCh38
rs1042514026 Chr.1: 172664244-172664244 on Build GRCh38
rs376636796 Chr.1: 172664415-172664415 on Build GRCh38
rs35774809 Chr.1: 172664355-172664355 on Build GRCh38
rs766758840 Chr.1: 172664406-172664406 on Build GRCh38
rs773493329 Chr.1: 172664401-172664401 on Build GRCh38
rs138441486 Chr.1: 172664365-172664365 on Build GRCh38
rs757862255 Chr.1: 172664348-172664348 on Build GRCh38
rs542772562 Chr.1: 172664289-172664289 on Build GRCh38
rs925281496 Chr.1: 172664286-172664286 on Build GRCh38
rs201525996 Chr.1: 172664397-172664397 on Build GRCh38
rs373975939 Chr.1: 172664325-172664325 on Build GRCh38
rs774626460 Chr.1: 172664376-172664376 on Build GRCh38
rs749273476 Chr.1: 172664362-172664362 on Build GRCh38
rs760897325 Chr.1: 172664405-172664405 on Build GRCh38
rs766811734 Chr.1: 172664340-172664340 on Build GRCh38
rs372021169 Chr.1: 172664398-172664398 on Build GRCh38
rs56168790 Chr.1: 172664251-172664251 on Build GRCh38
rs561262389 Chr.1: 172664352-172664352 on Build GRCh38
rs763631988 Chr.1: 172664349-172664349 on Build GRCh38
rs370579312 Chr.1: 172664319-172664319 on Build GRCh38

View Full Product Details