Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176568503-176568772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR2
Entrez Gene ID
54825
Gene Name
cadherin related family member 2
Gene Aliases
PCDH24, PCLKC
UniGene
Hs.4205
Gene Chromosome Location
Chr.5: 176542511-176595768 on Build GRCh38
Transcripts
NM_001171976.1 NM_017675.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3340385 c.155A>G p.D52G Chr.5: 176568708-176568708 on Build GRCh38
2154245 c.203A>C p.Y68S Chr.5: 176568756-176568756 on Build GRCh38
2154244 c.202T>C p.Y68H Chr.5: 176568755-176568755 on Build GRCh38
6587949 c.125-6C>T p.? Chr.5: 176568672-176568672 on Build GRCh38
4435030 c.216C>T p.V72V Chr.5: 176568769-176568769 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12716317 Chr.5: 176568570-176568570 on Build GRCh38
rs752803658 Chr.5: 176568742-176568742 on Build GRCh38
rs542492323 Chr.5: 176568590-176568590 on Build GRCh38
rs760410069 Chr.5: 176568658-176568658 on Build GRCh38
rs1020321480 Chr.5: 176568680-176568680 on Build GRCh38
rs1038108227 Chr.5: 176568616-176568616 on Build GRCh38
rs114185037 Chr.5: 176568710-176568710 on Build GRCh38
rs781376739 Chr.5: 176568652-176568652 on Build GRCh38
rs763921301 Chr.5: 176568715-176568715 on Build GRCh38
rs570719718 Chr.5: 176568743-176568743 on Build GRCh38
rs113573058 Chr.5: 176568741-176568741 on Build GRCh38
rs759138873 Chr.5: 176568702-176568702 on Build GRCh38
rs780131657 Chr.5: 176568630-176568630 on Build GRCh38
rs749604911 Chr.5: 176568687-176568687 on Build GRCh38
rs536943624 Chr.5: 176568591-176568591 on Build GRCh38
rs760224259 Chr.5: 176568713-176568713 on Build GRCh38
rs551092177 Chr.5: 176568553-176568553 on Build GRCh38
rs773818234 Chr.5: 176568683-176568683 on Build GRCh38
rs562503659 Chr.5: 176568563-176568563 on Build GRCh38
rs1046726718 Chr.5: 176568541-176568541 on Build GRCh38
rs765026569 Chr.5: 176568739-176568739 on Build GRCh38
rs1010490435 Chr.5: 176568667-176568667 on Build GRCh38
rs139206310 Chr.5: 176568709-176568709 on Build GRCh38
rs201557706 Chr.5: 176568639-176568639 on Build GRCh38
rs757131329 Chr.5: 176568627-176568627 on Build GRCh38
rs978795686 Chr.5: 176568714-176568714 on Build GRCh38
rs771395009 Chr.5: 176568695-176568695 on Build GRCh38
rs371036927 Chr.5: 176568660-176568660 on Build GRCh38
rs372721450 Chr.5: 176568657-176568657 on Build GRCh38
rs146915793 Chr.5: 176568703-176568703 on Build GRCh38
rs1030511252 Chr.5: 176568645-176568645 on Build GRCh38
rs755214796 Chr.5: 176568740-176568740 on Build GRCh38
rs766601111 Chr.5: 176568717-176568717 on Build GRCh38
rs747165658 Chr.5: 176568638-176568638 on Build GRCh38
rs113759967 Chr.5: 176568662-176568662 on Build GRCh38
rs1005909055 Chr.5: 176568608-176568608 on Build GRCh38
rs1037844353 Chr.5: 176568619-176568619 on Build GRCh38
rs770808217 Chr.5: 176568685-176568685 on Build GRCh38
rs774710686 Chr.5: 176568701-176568701 on Build GRCh38
rs886751111 Chr.5: 176568543-176568543 on Build GRCh38
rs917384712 Chr.5: 176568729-176568729 on Build GRCh38
rs748164417 Chr.5: 176568712-176568712 on Build GRCh38
rs770170201 Chr.5: 176568679-176568679 on Build GRCh38
rs150107118 Chr.5: 176568731-176568731 on Build GRCh38
rs949313164 Chr.5: 176568537-176568537 on Build GRCh38

View Full Product Details