Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57897332-57897595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGB1
Entrez Gene ID
1258
Gene Name
cyclic nucleotide gated channel beta 1
Gene Aliases
CNCG2, CNCG3L, CNCG4, CNG4, CNGB1B, GAR1, GARP, GARP2, RCNC2, RCNCb, RCNCbeta, RP45
UniGene
Hs.147062
Gene Chromosome Location
Chr.16: 57882340-57971116 on Build GRCh38
Transcripts
NM_001286130.1 NM_001297.4 XM_011522870.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6381580 c.3124C>A p.R1042S Chr.16: 57897515-57897515 on Build GRCh38
1679122 c.3148G>A p.G1050R Chr.16: 57897491-57897491 on Build GRCh38
319525 c.3239C>T p.A1080V Chr.16: 57897400-57897400 on Build GRCh38
6631177 c.3142G>A p.A1048T Chr.16: 57897497-57897497 on Build GRCh38
4061477 c.3199G>A p.V1067M Chr.16: 57897440-57897440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374172975 Chr.16: 57897409-57897409 on Build GRCh38
rs767619701 Chr.16: 57897529-57897529 on Build GRCh38
rs774960446 Chr.16: 57897426-57897426 on Build GRCh38
rs367764281 Chr.16: 57897459-57897459 on Build GRCh38
rs760310888 Chr.16: 57897390-57897391 on Build GRCh38
rs748180946 Chr.16: 57897510-57897510 on Build GRCh38
rs773593999 Chr.16: 57897454-57897454 on Build GRCh38
rs775022391 Chr.16: 57897380-57897380 on Build GRCh38
rs200287627 Chr.16: 57897505-57897505 on Build GRCh38
rs780864617 Chr.16: 57897540-57897540 on Build GRCh38
rs748288939 Chr.16: 57897569-57897569 on Build GRCh38
rs369503135 Chr.16: 57897507-57897507 on Build GRCh38
rs756012260 Chr.16: 57897535-57897535 on Build GRCh38
rs767959077 Chr.16: 57897447-57897447 on Build GRCh38
rs765306729 Chr.16: 57897554-57897557 on Build GRCh38
rs750087887 Chr.16: 57897573-57897573 on Build GRCh38
rs780388472 Chr.16: 57897503-57897503 on Build GRCh38
rs545721406 Chr.16: 57897417-57897417 on Build GRCh38
rs192178968 Chr.16: 57897517-57897517 on Build GRCh38
rs773698242 Chr.16: 57897514-57897514 on Build GRCh38
rs763490882 Chr.16: 57897478-57897478 on Build GRCh38
rs539304668 Chr.16: 57897504-57897504 on Build GRCh38
rs779688908 Chr.16: 57897541-57897541 on Build GRCh38
rs775265608 Chr.16: 57897518-57897518 on Build GRCh38
rs757969882 Chr.16: 57897476-57897476 on Build GRCh38
rs866891545 Chr.16: 57897375-57897375 on Build GRCh38
rs757167855 Chr.16: 57897371-57897371 on Build GRCh38
rs578138182 Chr.16: 57897491-57897491 on Build GRCh38
rs556751911 Chr.16: 57897396-57897396 on Build GRCh38
rs1049954541 Chr.16: 57897379-57897379 on Build GRCh38
rs148999583 Chr.16: 57897524-57897524 on Build GRCh38
rs755959274 Chr.16: 57897496-57897496 on Build GRCh38
rs752290959 Chr.16: 57897464-57897464 on Build GRCh38
rs535148864 Chr.16: 57897378-57897378 on Build GRCh38
rs767907011 Chr.16: 57897365-57897366 on Build GRCh38
rs371348182 Chr.16: 57897388-57897388 on Build GRCh38
rs200581517 Chr.16: 57897492-57897492 on Build GRCh38
rs897497044 Chr.16: 57897462-57897462 on Build GRCh38
rs779121632 Chr.16: 57897565-57897565 on Build GRCh38
rs200202348 Chr.16: 57897511-57897511 on Build GRCh38
rs142930143 Chr.16: 57897525-57897525 on Build GRCh38
rs9924712 Chr.16: 57897395-57897395 on Build GRCh38
rs143936383 Chr.16: 57897367-57897367 on Build GRCh38
rs201023939 Chr.16: 57897410-57897410 on Build GRCh38
rs751034406 Chr.16: 57897485-57897485 on Build GRCh38
rs753353134 Chr.16: 57897489-57897489 on Build GRCh38
rs531122937 Chr.16: 57897557-57897557 on Build GRCh38
rs761350175 Chr.16: 57897528-57897528 on Build GRCh38
rs750379218 Chr.16: 57897533-57897533 on Build GRCh38
rs760975936 Chr.16: 57897456-57897456 on Build GRCh38
rs763732525 Chr.16: 57897421-57897421 on Build GRCh38
rs756806434 Chr.16: 57897496-57897497 on Build GRCh38
rs753617403 Chr.16: 57897547-57897547 on Build GRCh38
rs762202033 Chr.16: 57897436-57897436 on Build GRCh38

View Full Product Details